Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2100493

Sigma-Aldrich

Anti-CSPG5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chondroitin sulfate proteoglycan 5 (neuroglycan C), Anti-MGC44034, Anti-NGC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CSPG5(10675)

Immunogen

Synthetic peptide directed towards the N terminal region of human CSPG5

Biochem/physiol Actions

CSPG5 may function as a growth and differentiation factor involved in neuritogenesis. It may induce ERBB3 activation.

Sequence

Synthetic peptide located within the following region: LGGVTAEAGSGDAQALPATLQAPHEVLGQSIMPPAIPEATEASGPPSPTP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shen Liu et al.
Molecular medicine reports, 20(5), 4059-4066 (2019-11-09)
The present study aimed to explore the role of the PTEN/Akt/mTOR signaling pathway in the neurite outgrowth and apoptosis of cortical neurons. Cortical neurons were seeded on or adjacent to chondroitin sulfate proteoglycans. The length, number and crossing behavior of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service