Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2100396

Sigma-Aldrich

Anti-CEBPA antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-C/EBP-α, Anti-CCAAT/enhancer binding protein (C/EBP), α, Anti-CEBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

37 kDa

species reactivity

, bovine, horse, rabbit, canine, pig, chicken, human, zebrafish, mouse, rat

technique(s)

western blot: suitable

immunogen sequence

RRERNNIAVRKSRDKAKQRNVETQQKVLELTSDNDRLRKRVEQLSRELDT

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEBPA(1050)

Immunogen

Peptide region of the protein sequence according to NP_004355.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lixiang Liu et al.
Adipocyte, 9(1), 279-289 (2020-06-25)
The quality and nutritional value of beef is closely linked to its content of intramuscular fat (IMF). The differentiation of preadipocytes and the deposition of lipid droplets in the adipocytes are the key to regulate the IMF content. The differentiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service