Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2100345

Sigma-Aldrich

Anti-CAV2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Caveolin 2, Anti-MGC12294

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

18 kDa

species reactivity

rat, sheep, horse, rabbit, guinea pig, human, mouse, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAV2(858)

General description

Caveolin 2 (CAV2) gene with three exons is mapped to human chromosome 7q31. The encoded protein is expressed in variety of human tissues, including the scleral spur cells, trabecular meshwork and retinal ganglion cells of the eye.

Immunogen

Synthetic peptide directed towards the N terminal region of human CAV2

Biochem/physiol Actions

Caveolin 2 (CAV2) along with CAV1, participates in transcytosis by inducing the formation of specialized invaginations of the plasma membrane called caveolae. CAV2 might be involved in the process of hormonal translocation. It is also implicated in the regulation of cancer progression. CAV2 plays an essential role in the regulation of sex hormone 17β-estradiol (E2)-dependent cellular proliferation. Genetic variation in the gene is associated with the increased risk of developing primary open-angle glaucoma (POAG).

Sequence

Synthetic peptide located within the following region: LVIPYNEKPEKPAKTQKTSLDEALQWRDSLDKLLQNNYGLASFKSFLKSE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Common variants near CAV1 and CAV2 are associated with primary open-angle glaucoma in Caucasians from the USA.
Wiggs JL
Human Molecular Genetics, 20(23), 4707-4713 (2011)
Caveolin proteins in signaling, oncogenic transformation and muscular dystrophy.
Razani B
Journal of Cell Science, 113 ( Pt 12), 2103-2109 (2000)
Clinical significance of Caveolin-1, Caveolin-2 and HER2/neu mRNA expression in human breast cancer.
Sagara Y
British Journal of Cancer, 91(5), 959-965 (2004)
Regulation of cancer cell proliferation by caveolin-2 down-regulation and re-expression.
Lee S
International Journal of Oncology, 38(5), 1395-1402 (2011)
Modulation of 17?-Estradiol Signaling on Cellular Proliferation by Caveolin-2.
Totta P
Journal of Cellular Physiology, 231(6), 1219-1225 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service