Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1413146

Sigma-Aldrich

Monoclonal Anti-LIPE (phospho S853) antibody produced in rabbit

clone FDO-12

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$418.00

$418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

antibody product type

primary antibodies

clone

FDO-12, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

western blot: 1:5000-1:10000

Storage temp.

−20°C

target post-translational modification

phosphorylation (pSer853)

Gene Information

human ... LIPE(3991)

Immunogen

A synthetic phosphopeptide corresponding to residues surrounding S853 of human LIPE.

Physical form

In PBS, 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service