Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1413018

Sigma-Aldrich

Monoclonal Anti-BTK (phospho Y223) antibody produced in rabbit

clone B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

antibody product type

primary antibodies

clone

B4, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

flow cytometry: 1-0.001 μg/mL

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr223)

Gene Information

human ... BTK(695)

Immunogen

A synthetic phosphopeptide corresponding to residues surrounding Y223 of human BTK.

Physical form

In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service