Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1412816

Sigma-Aldrich

Anti-NPTXR antibody produced in mouse

purified immunoglobulin

Synonym(s):

Anti-NPR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

form

buffered aqueous solution

mol wt

antigen 53.79 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NPTXR(23467)

General description

This gene encodes a protein similar to the rat neuronal pentraxin receptor. The rat pentraxin receptor is an integral membrane protein that is thought to mediate neuronal uptake of the snake venom toxin, taipoxin, and its transport into the synapses. Studies in rat indicate that translation of this mRNA initiates at a non-AUG (CUG) codon. This may also be true for mouse and human, based on strong sequence conservation amongst these species. (provided by RefSeq)

Immunogen

NPTXR (AAI53028.1, 1 a.a. ~ 489 a.a) full-length human protein.

Sequence
MLAFLGAVICIIASVPLAASPARALPGGADNASVASGAAASPGPQRSLSALHGAGGSAGPPALPGAPAASAHPLPPGPLFSRFLCTPLAAACPSGAQQGDAAGAAPGEREELLLLQSTAEQLRQTALQQEARIRADQDTIRELTGKLGRCESGLPRGLQGAGPRRDTMADGPWDSPALILELEDAVRALRDRIDRLEQELPARVNLSAAPAPVSAVPTGLHSKMDQLEGQLLAQVLALEKERVALSHSSRRQRQEVEKELDVLQGRVAELEHGSSAYSPPDAFKISIPIRNNYMYARVRKALPELYAFTACMWLRSRSSGTGQGTPFSYSVPGQANEIVLLEAGHEPMELLINDKVAQLPLSLKDNGWHHICIAWTTRDGLWSAYQDGELQGSGENLAAWHPIKPHGILILGQEQDTLGGRFDATQAFVGDIAQFNLWDHALTPAQVLGIANCTAPLLGNVLPWEDKLVEAFGGATKAAFDVCKGRAKA

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service