Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1412040

Sigma-Aldrich

ANTI-ISL1 antibody produced in mouse

clone 8A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ISL1, Isl-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8A3, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.41 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ISL1(3670)

General description

Insulin gene enhancer binding protein-1 (ISL-1) gene spanning 220kb of genomic DNA, is mapped to human chromosome 5q11.1. The gene codes for a member of LIM zinc-binding/homeobox-domain transcription factor and is predominantly expressed in hindbrain, forebrain, ventral neural tube, dorsal root ganglia (DRG) and trigeminal ganglia.

Immunogen

ISL1 (NP_002193, 63 a.a. ~ 159 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YCKRDYIRLYGIKCAKCSIGFSKNDFVMRARSKVYHIECFRCVACSRQLIPGDEFALREDGLFCRADHDVVERASLGAGDPLSPLHPARPLQMAAEP

Biochem/physiol Actions

Insulin gene enhancer binding protein-1 (ISL-1) is a transcription factor which plays a vital role in the development of the heart, motor neurons and pancreas. It also stimulates adult pancreatic ß-cells proliferation. ISL1 might also be involved in the regulation of insulin expression. Elevated expression of the gene has been observed in patients with gastric cancer. Thus, the protein can be used as a potential biomarker and therapeutic target for gastric cancer. Mutation in the gene is associated with the increased risk of susceptibility to human congenital heart disease (CHD).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study and meta-analysis identify ISL1 as genome-wide significant susceptibility gene for bladder exstrophy
Draaken M
PLoS Genetics, 11 (2015)
Common variation in ISL1 confers genetic susceptibility for human congenital heart disease.
Stevens KN
PLoS ONE, 5 (2010)
An abnormally high expression of ISL-1 represents a potential prognostic factor in gastric cancer.
Guo C
Human Pathology, 46, 1282-1289 (2015)
A family of LIM domain-associated cofactors confer transcriptional synergism between LIM and Otx homeodomain proteins.
Bach I
Genes & Development, 11, 1370-1380 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service