Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411932

Sigma-Aldrich

ANTI-ZKSCAN3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ33906, KIAA0426, ZF47, ZFP306, ZKSCAN3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZKSCAN3(80317)

General description

Zinc finger with KRAB and SCAN domains 3 (ZKSCAN3) is encoded by the gene mapped to human chromosome 6p22.1. The encoded protein belongs to the zinc finger family of DNA-binding proteins.

Immunogen

ZKSCAN3 (NP_077819.1, 1 a.a. ~ 538 a.a) full-length human protein.

Sequence
MARELSESTALDAQSTEDQMELLVIKVEEEEAGFPSSPDLGSEGSRERFRGFRYPEAAGPREALSRLRELCRQWLQPEMHSKEQILELLVLEQFLTILPGNLQSWVREQHPESGEEVVVLLEYLERQLDEPAPQVSGVDQGQELLCCKMALLTPAPGSQSSQFQLMKALLKHESVGSQPLQDRVLQVPMLAHGGCCREDAVVASRLTPESQGLLKVEDVALTLTPEWTQQDSSQGNLCRDEKQENHGSLVSLGDEKQTKSRDLPPAEELPEKEHGKISCHLREDIAQIPTCAEAGEQEGRLQRKQKNATGGRRHICHECGKSFAQSSGLSKHRRIHTGEKPYECEECGKAFIGSSALVIHQRVHTGEKPYECEECGKAFSHSSDLIKHQRTHTGEKPYECDDCGKTFSQSCSLLEHHRIHTGEKPYQCSMCGKAFRRSSHLLRHQRIHTGDKNVQEPEQGEAWKSRMESQLENVETPMSYKCNECERSFTQNTGLIEHQKIHTGEKPYQCNACGKGFTRISYLVQHQRSHVGKNILSQ

Biochem/physiol Actions

Zinc finger with KRAB and SCAN domains 3 (ZKSCAN3) acts as a zinc finger transcription factor and plays an essential role in stimulating prostate cancer cell migration. Elevated expression of the gene is associated with various types of cancers, including prostate cancer, colon tumors, colorectal cancer (CRC) and multiple myeloma (MM). The encoded protein functions as a chief transcriptional repressor of autophagy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Yang et al.
Oncogene, 30(11), 1329-1340 (2010-11-09)
Dysregulation of cyclin D2 contributes to the pathogenesis of multiple myeloma, and can occur through translocations that activate MAF/MAFB or MMSET/FGFR3. However, cyclin D2 induction can also be seen in the absence of such translocations, such as in patients with
Santosh Chauhan et al.
Molecular cell, 50(1), 16-28 (2013-02-26)
Autophagy constitutes a major cell-protective mechanism that eliminates damaged components and maintains energy homeostasis via recycling nutrients under normal/stressed conditions. Although the core components of autophagy have been well studied, regulation of autophagy at the transcriptional level is poorly understood.
Xingding Zhang et al.
The international journal of biochemistry & cell biology, 44(7), 1166-1173 (2012-04-26)
In our previous studies, ZKSCAN3 was demonstrated to be over-expressed in invasive colonic tumor cells and their liver metastases, but minimally expressed in adjacent non-transformed tissues. Further preliminary data showed that ZKSCAN3 was expressed in a majority of prostate cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service