Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411904

Sigma-Aldrich

ANTI-PCDHGA11 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

PCDH-GAMMA-A11, PCDHGA11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 91.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. (provided by RefSeq)

Immunogen

PCDHGA11 (NP_114480.1, 1 a.a. ~ 837 a.a) full-length human protein.

Sequence
MANRLQRGDRSRLLLLLCIFLGTLRGFRARQIRYSVPEETEKGSFVGNISKDLGLEPRELAKRGVRIVSRGKTQLFAVNPRSGSLITAGRIDREELCETVSSCFLNMELLVEDTLKIYGVEVEIIDINDNAPSFQEDEVEIKVSEHAIPGARFALPNARDPDVGVNSLQSYQLSPNNYFSLQLRGRTDGAKNPELVLEGSLDREKEAAHLLLLTALDGGDPIRKGAVPIRVVVLDVNDHIPMFTQSVYRVSVPENISSGTRVLMVNATDPDEGINGEVMYSFRNMESKASEIFQLDSQTGEVQVRGSLDFEKYRFYEMEIQGQDGGGLFTTTTMLITVVDVNDNAPEITITSSINSILENSPPGTVIALLNVQDQDSGENGQVSCFIPNHLPFKLEKTYGNYYKLITSRVLDRELVQSYNITLTATDQGSPPLSAETHVWLNVADDNDNPPVFPHSSYSAYIPENNPRGASIFSVTALDPDSKQNALVTYSLTDDTVQGVPLSSYVSINSNTGVLYALQSFDYEQFRDLELRVIARDSGDPPLSSNVSLSLFVLDQNDNAPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDKDSGQNAWLSYRLLKASEPGLFAVGEHTGEVRTARALLDRDALKQSLVVAVQDHGQPPLSATVTLTVAVADSIPEVLADLGSLESLANSETSDLSLYLVVAVAAVSCIFLVFVIVLLALRLWRWHKSRLLQASEGGLAGMPTSHFVGVDGVQAFLQTYSHEVSLIADSQKSHLIFPQPNYGDTLISQESCEKSEPLLIAEDSAIILGKCDPTSNQVRFISLPPNCWCLGTSLLRRCFLSLL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service