Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1411872

Sigma-Aldrich

ANTI-PTCH antibody produced in mouse

clone 4F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

BCNS, FLJ26746, FLJ42602, HPE7, PTCH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F11, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.74 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTCH(5727)

General description

This gene encodes a member of the patched gene family. The encoded protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis, as well as the desert hedgehog and indian hedgehog proteins. This gene functions as a tumor suppressor. Mutations of this gene have been associated with Gorlin Syndrome(BCNS), esophageal squamous cell carcinoma, trichoepitheliomas, transitional cell carcinomas of the bladder, as well as holoprosencephaly. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional splice variants have been described, but their full length sequences and biological validity cannot be determined currently. (provided by RefSeq)

Immunogen

PTCH (NP_000255, 651 a.a. ~ 750 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
AHETQITMQSTVQLRTEYDPHTHVYYTTAEPRSEISVQPVTVTQDTLSCQSPESTSSTRDLLSQFSDSSLHCLEPPCTKWTLSSFAEKHYAPFLLKPKAK*

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service