SAB1411846
ANTI-ADIPOQ antibody produced in mouse
clone 3H2, purified immunoglobulin, buffered aqueous solution
Synonym(s):
ACDC, ACRP30, ADIPOQ, ADPN, APM-1
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
3H2, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable
isotype
IgG2bκ
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... ADIPOQ(9370)
Immunogen
Mouse monoclonal antibody raised against adiponectin.
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service