Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SAB1411419

Sigma-Aldrich

Anti-OTUB2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

C14orf137, FLJ21916, MGC3102, OTB2, OTU2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27.2 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OTUB2(78990)

General description

OTU ubiquitin aldehyde binding 2 (OTUB2) is a de-ubiquitinating enzyme, which belongs to the ovarian tumor protease (OTU) subfamily of deubiquitinylases. It has an ovarian tumor (OTU) domain. OTUB2 gene is mapped to human chromosome 14q32.

Immunogen

OTUB2 (NP_075601.1, 1 a.a. ~ 234 a.a) full-length human protein.

Sequence
MSETSFNLISEKCDILSILRDHPENRIYRRKIEELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNFFNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDFCTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNILYAADKH

Biochem/physiol Actions

OTU ubiquitin aldehyde binding 2 (OTUB2) possesses deubiquitinating enzymes (DUB) activity. It is capable of cleaving polyubiquitin chains, that are differently linked. It plays an important role in regulating the degradation of GLI family zinc finger 2 (Gli2) protein. OTUB2 can inhibit the death of human β cells. It is involved in immunity and viral infection. This protein can modulate nuclear factor κB (NF-κB) signaling pathway. OTUB2 might be associated with endometrial cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol A Edwards et al.
BMC evolutionary biology, 7, 157-157 (2007-09-08)
The evolution of genomic imprinting, the parental-origin specific expression of genes, is the subject of much debate. There are several theories to account for how the mechanism evolved including the hypothesis that it was driven by the evolution of X-inactivation
Dakshinamurthy Sivakumar et al.
The Journal of biological chemistry, 295(20), 6972-6982 (2020-04-09)
The ovarian tumor domain (OTU) deubiquitinylating cysteine proteases OTUB1 and OTUB2 (OTU ubiquitin aldehyde binding 1 and 2) are representative members of the OTU subfamily of deubiquitinylases. Deubiquitinylation critically regulates a multitude of important cellular processes, such as apoptosis, cell
A Beck et al.
Diabetologia, 56(6), 1317-1326 (2013-03-22)
Pro-inflammatory cytokines induce death of beta cells and hamper engraftment of transplanted islet mass. Our aim was to reveal novel genes involved in this process, as a platform for innovative therapeutic approaches. Small interfering RNA (siRNA) high-throughput screening (HTS) of
Xin-Yan Li et al.
Biochemical and biophysical research communications, 505(1), 113-118 (2018-09-23)
The transcription factor Gli2 plays crucial roles in the transduction of Hedgehog (Hh) signals, yet the mechanisms that control Gli2 degradation remain unclear. Here we have identified the eubiquitinating enzyme otubain2 (OTUB2) as a regulator of Gli2 protein degradation. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service