Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1411396

Sigma-Aldrich

Anti-WNT5A antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

hWNT5A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42.3 kDa

species reactivity

human

technique(s)

proximity ligation assay: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNT5A(7474)

General description

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 98%, 98% and 87% amino acid identity to the mouse, rat and the xenopus Wnt5A protein, respectively. The experiments performed in Xenopus laevis embryos identified that human frizzled-5 (hFz5) is the receptor for the Wnt5A ligand and the Wnt5A/hFz5 signaling mediates axis induction. (provided by RefSeq)
Wnt family member 5A (WNT5A) is a secreted, proinflammatory protein. It belongs to the noncanonical WNT family. The gene encoding this protein is mapped to human chromosome 3p14.3.

Immunogen

WNT5A (NP_003383.2, 1 a.a. ~ 380 a.a) full-length human protein.

Sequence
MKKSIGILSPGVALGMAGSAMSSKFFLVALAIFFSFAQVVIEANSWWSLGMNNPVQMSEVYIIGAQPLCSQLAGLSQGQKKLCHLYQDHMQYIGEGAKTGIKECQYQFRHRRWNCSTVDNTSVFGRVMQIGSRETAFTYAVSAAGVVNAMSRACREGELSTCGCSRAARPKDLPRDWLWGGCGDNIDYGYRFAKEFVDARERERIHAKGSYESARILMNLHNNEAGRRTVYNLADVACKCHGVSGSCSLKTCWLQLADFRKVGDALKEKYDSAAAMRLNSRGKLVQVNSRFNSPTTQDLVYIDPSPDYCVRNESTGSLGTQGRLCNKTSEGMDGCELMCCGRGYDQFKTVQTERCHCKFHWCCYVKCKKCTEIVDQFVCK

Biochem/physiol Actions

Wnt family member 5A (WNT5A) has a role in metabolic dysfunction in obesity and negatively modulates angiogenesis in adipose tissue. The protein regulates cell motility and polarity by activating β-catenin–independent pathways. It has been shown to be overexpressed in gliomas. In addition, elevated expression of Wnt5a also contributes to the development of human melanoma cells. Thus, Wnt5a pathway is suggested to be a potential target for the treatment of melanoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional mechanisms of WNT5A based on NF-?B, Hedgehog, TGF?, and Notch signaling cascades
Masuko Katoh
International Journal of Molecular Medicine, 23(6), 763-769 (2009)
Ashani T Weeraratna et al.
Cancer cell, 1(3), 279-288 (2002-06-28)
Gene expression profiling identified human melanoma cells demonstrating increased cell motility and invasiveness. The gene WNT5A best determined in vitro invasive behavior. Melanoma cells were transfected with vectors constitutively overexpressing Wnt5a. Consistent changes included actin reorganization and increased cell adhesion.
Shakun Karki et al.
American journal of physiology. Heart and circulatory physiology, 313(1), H200-H206 (2017-04-16)
Experimental studies have suggested that Wingless-related integration site 5A (WNT5A) is a proinflammatory secreted protein that is associated with metabolic dysfunction in obesity. Impaired angiogenesis in fat depots has been implicated in the development of adipose tissue capillary rarefaction, hypoxia
Elena Binda et al.
Cancer research, 77(4), 996-1007 (2016-12-25)
Brain invasion by glioblastoma determines prognosis, recurrence, and lethality in patients, but no master factor coordinating the invasive properties of glioblastoma has been identified. Here we report evidence favoring such a role for the noncanonical WNT family member Wnt5a. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service