Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1411265

Sigma-Aldrich

Anti-SGSH antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HSS, MPS3A, SFMD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SGSH(6448)

General description

This gene encodes one of several enzymes involved in the lysosomal degradation of heparan sulfate. Mutations in this gene are associated with Sanfilippo syndrome A, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. Transcripts of varying sizes have been reported but their biological validity has not been determined. (provided by RefSeq)

Immunogen

SGSH (NP_000190.1, 1 a.a. ~ 502 a.a) full-length human protein.

Sequence
MSCPVPACCALLLVLGLCRARPRNALLLLADDGGFESGAYNNSAIATPHLDALARRSLLFRNAFTSVSSCSPSRASLLTGLPQHQNGMYGLHQDVHHFNSFDKVRSLPLLLSQAGVRTGIIGKKHVGPETVYPFDFAYTEENGSVLQVGRNITRIKLLVRKFLQTQDDRPFFLYVAFHDPHRCGHSQPQYGTFCEKFGNGESGMGRIPDWTPQAYDPLDVLVPYFVPNTPAARADLAAQYTTVGRMDQGVGLVLQELRDAGVLNDTLVIFTSDNGIPFPSGRTNLYWPGTAEPLLVSSPEHPKRWGQVSEAYVSLLDLTPTILDWFSIPYPSYAIFGSKTIHLTGRSLLPALEAEPLWATVFGSQSHHEVTMSYPMRSVQHRHFRLVHNLNFKMPFPIDQDFYVSPTFQDLLNRTTAGQPTGWYKDLRHYYYRARWELYDRSRDPHETQNLATDPRFAQLLEMLRDQLAKWQWETHDPWVCAPDGVLEEKLSPQCQPLHNEL

Biochem/physiol Actions

The gene SGSH (N-sulfoglucosamine sulfohydrolase) encodes an enzyme that catalyzes the hydrolysis of N-linked sulfate groups from the GAGs (glycosaminoglycans) heparan sulfate and heparin. Deficiency of this enzyme due to mutations causes lysosomal storage of glyco­aminoglycans. This leads to a disease called the mucopolysaccharidosis type IIIA or Sanfilippo A syndrome, which is a childhood-onset neurodegenerative disease characterized by facial, visceral and skeletal abnormalities.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weichun Xiao et al.
American journal of physiology. Cell physiology, 309(4), C215-C227 (2015-06-26)
Nonalcoholic steatohepatitis (NASH) is the progressive form of nonalcoholic fatty liver disease and so far is supposed to be related with mitochondrial impairment. Hepatic stimulator substance (HSS) has been defined as a liver-protective factor promoting hepatocyte DNA synthesis and hepatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service