Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1411149

Sigma-Aldrich

Anti-TWF1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

A6, MGC23788, MGC41876, PTK9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43.9 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TWF1(5756)

General description

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. (provided by RefSeq)

Immunogen

TWF1 (NP_002813.2, 1 a.a. ~ 384 a.a) full-length human protein.

Sequence
MTRRGRRRSSHFLGPPAGAAGCTQRRSRELAAAAMSHQTGIQASEDVKEIFARARNGKYRLLKISIENEQLVIGSYSQPSDSWDKDYDSFVLPLLEDKQPCYILFRLDSQNAQGYEWIFIAWSPDHSHVRQKMLYAATRATLKKEFGGGHIKDEVFGTVKEDVSLHGYKKYLLSQSSPAPLTAAEEELRQIKINEVQTDVGVDTKHQTLQGVAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSFAKPKGPAGKRGIRRLIRGPAETEATTD

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service