Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411004

Sigma-Aldrich

Anti-DDX56 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DDX21, DDX26, NOH61

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX56(54606)

General description

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit. (provided by RefSeq)

Immunogen

DDX56 (NP_061955.1, 1 a.a. ~ 547 a.a) full-length human protein.

Sequence
MEDSEALGFEHMGLDPRLLQAVTDLGWSRPTLIQEKAIPLALEGKDLLARARTGSGKTAAYAIPMLQLLLHRKATGPVVEQAVRGLVLVPTKELARQAQSMIQQLATYCARDVRVANVSAAEDSVSQRAVLMEKPDVVVGTPSRILSHLQQDSLKLRDSLELLVVDEADLLFSFGFEEELKSLLCHLPRIYQAFLMSATFNEDVQALKELILHNPVTLKLQESQLPGPDQLQQFQVVCETEEDKFLLLYALLKLSLIRGKSLLFVNTLERSYRLRLFLEQFSIPTCVLNGELPLRSRCHIISQFNQGFYDCVIATDAEVLGAPVKGKRRGRGPKGDKASDPEAGVARGIDFHHVSAVLNFDLPPTPEAYIHRAGRTARANNPGIVLTFVLPTEQFHLGKIEELLSGENRGPILLPYQFRMEEIEGFRYRCRDAMRSVTKQAIREARLKEIKEELLHSEKLKTYFEDNPRDLQLLRHDLPLHPAVVKPHLGHVPDYLVPPALRGLVRPHKKRKKLSSSCRKAKRAKSQNPLRSFKHKGKKFRPTAKPS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service