Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1410976

Sigma-Aldrich

Anti-PLOD2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

LH2, TLH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87.1 kDa

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PLOD2(5352)

General description

The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

PLOD2 (NP_891988.1, 1 a.a. ~ 758 a.a) full-length human protein.

Sequence
MGGCTVKPQLLLLALVLHPWNPCLGADSEKPSSIPTDKLLVITVATKESDGFHRFMQSAKYFNYTVKVLGQGEEWRGGDGINSIGGGQKVRLMKEVMEHYADQDDLVVMFTECFDVIFAGGPEEVLKKFQKANHKVVFAADGILWPDKRLADKYPVVHIGKRYLNSGGFIGYAPYVNRIVQQWNLQDNDDDQLFYTKVYIDPLKREAINITLDHKCKIFQTLNGAVDEVVLKFENGKARAKNTFYETLPVAINGNGPTKILLNYFGNYVPNSWTQDNGCTLCEFDTVDLSAVDVHPNVSIGVFIEQPTPFLPRFLDILLTLDYPKEALKLFIHNKEVYHEKDIKVFFDKAKHEIKTIKIVGPEENLSQAEARNMGMDFCRQDEKCDYYFSVDADVVLTNPRTLKILIEQNRKIIAPLVTRHGKLWSNFWGALSPDGYYARSEDYVDIVQGNRVGVWNVPYMANVYLIKGKTLRSEMNERNYFVRDKLDPDMALCRNAREMTLQREKDSPTPETFQMLSPPKGVFMYISNRHEFGRLLSTANYNTSHYNNDLWQIFENPVDWKEKYINRDYSKIFTENIVEQPCPDVFWFPIFSEKACDELVEEMEHYGKWSGGKHHDSRISGGYENVPTDDIHMKQVDLENVWLHFIREFIAPVTLKVFAGYYTKGFALLNFVVKYSPERQRSLRPHHDASTFTINIALNNVGEDFQGGGCKFLRYNCSIESPRKGWSFMHPGRLTHLHEGLPVKNGTRYIAVSFIDP

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service