Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410898

Sigma-Aldrich

Anti-MECR antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CGI-63, FASN2B, NRBF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 40.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MECR(51102)

General description

MECR (Mitochondrial trans-2-enoyl-CoA reductase) is a mitochondrial localized protein, also referred to as nuclear receptor binding factor-1 (NRBF-1). It consists of N-terminal mitochondrial targeting sequence, which plays an essential role for its mitochondrial localization. It was firstly identified as a binding protein of the nuclear hormone receptor peroxisome proliferator-activated receptor (PPARα).

Immunogen

MECR (NP_057095.2, 1 a.a. ~ 373 a.a) full-length human protein.

Sequence
MWVCSTLWRVRTPARQWRGLLPASGCHGPAASSYSASAEPARVRALVYGHHGDPAKVVELKNLELAAVRGSDVRVKMLAAPINPSDINMIQGNYGFLPELPAVGGNEGVAQVVAVGSNVTGLKPGDWVIPANAGLGTWRTEAVFSEEALIQVPSDIPLQSAATLGVNPCTAYRMLMDFEQLQPGDSVIQNASNSGVGQAVIQIAAALGLRTINVVRDRPDIQKLSDRLKSLGAEHVITEEELRRPEMKNFFKDMPQPRLALNCVGGKSSTELLRQLARGGTMVTYGGMAKQPVVASVSLLIFKDLKLRGFWLSQWKKDHSPDQFKELILTLCDLIRRGQLTAPACSQVPLQDYQSALEASMKPFISSKQILTM

Biochem/physiol Actions

MECR (Mitochondrial trans-2-enoyl-CoA reductase) plays a vital role in the intracellular signaling pathways of fatty acid elongation cycle pathway. In the nuclei, it catalyzes reduction reaction of trans-2-enoyl thioesters to the corresponding saturated acyl thioesters in the presence of NADPH. It has also been reported that MECR acts as a component of the mitochondrial FAS II (fatty acid synthesis II) pathway and enhances the PPARa-dependent luciferase activity in HeLa cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Gyu Kim et al.
Endocrinology and metabolism (Seoul, Korea), 29(2), 185-194 (2014-07-18)
Mitochondrial trans-2-enoyl-CoA reductase (MECR) is involved in mitochondrial synthesis of fatty acids and is highly expressed in mitochondria. MECR is also known as nuclear receptor binding factor-1, which was originally reported with yeast two-hybrid screening as a binding protein of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service