Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1410482

Sigma-Aldrich

Anti-CHRDL2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

BNF1, CHL2, DKFZp586N2124, FKSG37

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49.61 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRDL2(25884)

Related Categories

General description

CHRDL2 (Chordin-like 2) is a novel chordin-related gene belonging to the chordin family of proteins. It encodes a putative extracellular matrix protein consistsing of a signal peptide and three cysteine-rich (CR) domains. In humans, it expressed in several normal adult tissues predominantly in liver, heart, prostate, testis, and ovary.

Immunogen

CHRDL2 (AAI53101.1, 1 a.a. ~ 451 a.a) full-length human protein.

Sequence
MVPEVRVLSSLLGLALLWFPLDSHARARPDMFCLFHGKRYSPGESWHPYLEPQGLMYCLRCTCSEGAHVSCYRLHCPPVHCPQPVTEPQQCCPKCVEPHTPSGLRAPPKSCQHNGTMYQHGEIFSAHELFPSRLPNQCVLCSCTEGQIYCGLTTCPEPGCPAPLPLPDSCCQACKDEASEQSDEEDSVQSLHGVRHPQDPCSSDAGRKRGPGTPAPTGLSAPLSFIPRHFRPKGAGSTTVKIVLKEKHKKACVHGGKTYSHGEVWHPAFRAFGPLPCILCTCEDGRQDCQRVTCPTEYPCRHPEKVAGKCCKICPEDKADPGHSEISSTRCPKAPGRVLVHTSVSPSPDNLRRFALEHEASDLVEIYLWKLVKDEETEAQRGEVPGPRPHSQNLPLDSDQESQEARLPERGTALPTARWPPRRSLERLPSPDPGAEGHGQSRQSDQDITKT

Biochem/physiol Actions

CHRDL2 (Chordin-like 2) may play a role in the cartilage regeneration, myoblast and osteoblast differentiation. It has been reported that CHRDL2 may negatively regulate the cartilage formation/regeneration from immature mesenchymal cells. CHRDL2 restricts BMPs activity by blocking their interaction with its type I BMP receptor ectodomain. Overexpressed expression of CHRDL2 has been reported in several cancerous tissues such as breast, lung and colon cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anat Oren et al.
Gene, 331, 17-31 (2004-04-20)
Chordin-like cysteine-rich repeats (CRs) are conserved domains present in an expanding family of secreted proteins that associate with members of the TGF beta superfamily. In this study, we report the molecular cloning and characterization of CHL2 (chordin-like 2), a novel
Inmin Wu et al.
Gene, 311, 105-110 (2003-07-11)
In an effort to identify novel genes relevant to tumor angiogenesis, we compared the genes expressed in a matched pair composed of vascularized breast tumor and its adjacent normal tissue obtained from the same cancer patient. Using differential display, we
Naoki Nakayama et al.
Development (Cambridge, England), 131(1), 229-240 (2003-12-09)
We have identified a novel chordin-like protein, CHL2, which is structurally most homologous to CHL/neuralin/ventroptin. When injected into Xenopus embryos, CHL2 RNA induced a secondary axis. Recombinant CHL2 protein interacted directly with BMPs in a competitive manner to prevent binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service