Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1410418

Sigma-Aldrich

Anti-VPS39 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ21681, FLJ46546, KIAA0770, TLP, VAM6, hVam6p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 100.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... VPS39(23339)

General description

This gene encodes a protein that may promote clustering and fusion of late endosomes and lysosomes. The protein may also act as an adaptor protein that modulates the transforming growth factor-beta response by coupling the transforming growth factor-beta receptor complex to the Smad pathway. (provided by RefSeq)

Immunogen

VPS39 (NP_056104.2, 1 a.a. ~ 875 a.a) full-length human protein.

Sequence
MHDAFEPVPILEKLPLQIDCLAAWEEWLLVGTKQGHLLLYRIRKDVGCNRFEVTLEKSNKNFSKKIQQIHVVSQFKILVSLLENNIYVHDLLTFQQITTVSKAKGASLFTCDLQHTETGEEVLRMCVAVKKKLQLYFWKDREFHELQGDFSVPDVPKSMAWCENSICVGFKRDYYLIRVDGKGSIKELFPTGKQLEPLVAPLADGKVAVGQDDLTVVLNEEGICTQKCALNWTDIPVAMEHQPPYIIAVLPRYVEIRTFEPRLLVQSIELQRPRFITSGGSNIIYVASNHFVWRLIPVPMATQIQQLLQDKQFELALQLAEMKDDSDSEKQQQIHHIKNLYAFNLFCQKRFDESMQVFAKLGTDPTHVMGLYPDLLPTDYRKQLQYPNPLPVLSGAELEKAHLALIDYLTQKRSQLVKKLNDSDHQSSTSPLMEGTPTIKSKKKLLQIIDTTLLKCYLHTNVALVAPLLRLENNHCHIEESEHVLKKAHKYSELIILYEKKGLHEKALQVLVDQSKKANSPLKGHERTVQYLQHLGTENLHLIFSYSVWVLRDFPEDGLKIFTEDLPEVESLPRDRVLGFLIENFKGLAIPYLEHIIHVWEETGSRFHNCLIQLYCEKVQGLMKEYLLSFPAGKTPVPAGEEEGELGEYRQKLLMFLEISSYYDPGRLICDFPFDGLLEERALLLGRMGKHEQALFIYVHILKDTRMAEEYCHKHYDRNKDGNKDVYLSLLRMYLSPPSIHCLGPIKLELLEPKANLQAALQVLELHHSKLDTTKALNLLPANTQINDIRIFLEKVLEENAQKKRFNQVLKNLLHAEFLRVQEERILHQQVKCIITEEKVCMVCKKKIGNSAFARYPNGVVVHYFCSKEVNPADT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service