Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1410200

Sigma-Aldrich

Anti-RASGRP4 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 74.9 kDa

species reactivity

human

technique(s)

proximity ligation assay: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The protein encoded by this gene is a member of the Ras guanyl nucleotide-releasing protein (RasGRP) family of Ras guanine nucleotide exchange factors. It contains a Ras exchange motif, a diacylglycerol-binding domain, and two calcium-binding EF hands. This protein was shown to activate H-Ras in a cation-dependent manner in vitro. Expression of this protein in myeloid cell lines was found to be correlated with elevated level of activated RAS protein, and the RAS activation can be greatly enhanced by phorbol ester treatment, which suggested a role of this protein in diacylglycerol regulated cell signaling pathways. Studies of a mast cell leukemia cell line expressing substantial amounts of abnormal transcripts of this gene indicated that this gene may play an important role in the final stages of mast cell development. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

RASGRP4 (NP_733749.1, 1 a.a. ~ 673 a.a) full-length human protein.

Sequence
MNRKDSKRKSHQECTGKIGGRGRPRQVRRHKTCPSPREISKVMASMNLGLLSEGGCSEDELLEKCIQSFDSAGSLCHEDHMLNMVLAMHSWVLPSADLAARLLTSYQKATGDTQELRRLQICHLVRYWLMRHPEVMHQDPQLEEVIGRFWATVAREGNSAQRRLGDSSDLLSPGGPGPPLPMSSPGLGKKRKVSLLFDHLETGELAQHLTYLEFRSFQAITPQDLRSYVLQGSVRGCPALEGSVGLSNSVSRWVQVMVLSRPGPLQRAQVLDKFIHVAQRLHQLQNFNTLMAVTGGLCHSAISRLKDSHAHLSPDSTKALLELTELLASHNNYARYRRTWAGCAGFRLPVLGVHLKDLVSLHEAQPDRLPDGRLHLPKLNNLYLRLQELVALQGQHPPCSANEDLLHLLTLSLDLFYTEDEIYELSYAREPRCPKSLPPSPFNAPLVVEWAPGVTPKPDRVTLGRHVEQLVESVFKNYDPEGRGTISQEDFERLSGNFPFACHGLHPPPRQGRGSFSREELTGYLLRASAICSKLGLAFLHTFHEVTFRKPTFCDSCSGFLWGVTKQGYRCRECGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service