Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1410020

Sigma-Aldrich

Monoclonal Anti-ARHGEF10 antibody produced in mouse

clone 6G5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686H0726, GEF10, MGC131664

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G5, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF10(9639)

General description

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. (provided by RefSeq)

Immunogen

ARHGEF10 (NP_055444, 792 a.a. ~ 890 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GKQDKSGRPTFFTAVFNTFTPAIKESWVNSLQMAKLALEEENHMGWFCVEDDGNHIKKEKHPLLVGHMPVMVAKQQEFKIECAAYNPEPYLNNESQPDS

Biochem/physiol Actions

ARHGEF10 (Rho guanine nucleotide exchange factor 10) works as a guanine nucleotide exchange factor for RhoA, RhoB and RhoC. It is involved in atherosclerotic cerebral infarction and mutation in ARHGEF10 is associated with increased risk of atherothrombotic stroke. Mutation in ARFGEF10 is also linked with slowed nerve conduction velocities and thin myelination of peripheral nerves.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Ying Yin et al.
Neurology India, 59(3), 408-412 (2011-07-12)
The gene encoding RhoA guanine nucleotide exchange factor 10(ARHGEF10) has been reported to be a risk factor for atherothrombotic stroke (AS) in Japanese. The single-nucleotide polymorphism (SNP) rs4376531 in intron 16 on ARHGEF10 is associated with AS and may play
Taro Chaya et al.
The Journal of biological chemistry, 286(34), 29511-29520 (2011-07-02)
The T332I mutation in Rho guanine nucleotide exchange factor 10 (ARHGEF10) was previously found in persons with slowed nerve conduction velocities and thin myelination of peripheral nerves. However, the molecular and cellular basis of the T332I mutant is not understood.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service