Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1409997

Sigma-Aldrich

Anti-MMP20 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MMP-20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53.13 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP20(9313)

General description

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene degrades amelogenin, the major protein component of dental enamel matrix, and so the protein is thought to play a role in tooth enamel formation. A mutation in this gene, which alters the normal splice pattern and results in premature termination of the encoded protein, has been associated with amelogenesis imperfecta. This gene is part of a cluster of MMP genes that localizes to chromosome 11q22.3. (provided by RefSeq)

Immunogen

MMP20 (NP_004762.2, 1 a.a. ~ 483 a.a) full-length human protein.

Sequence
MKVLPASGLAVFLIMALKFSTAAPSLVAASPRTWRNNYRLAQAYLDKYYTNKEGHQIGEMVARGSNSMIRKIKELQAFFGLQVTGKLDQTTMNVIKKPRCGVPDVANYRLFPGEPKWKKNTLTYRISKYTPSMSSVEVDKAVEMALQAWSSAVPLSFVRINSGEADIMISFENGDHGDSYPFDGPRGTLAHAFAPGEGLGGDTHFDNAEKWTMGTNGFNLFTVAAHEFGHALGLAHSTDPSALMYPTYKYKNPYGFHLPKDDVKGIQALYGPRKVFLGKPTLPHAPHHKPSIPDLCDSSSSFDAVTMLGKELLLFKDRIFWRRQVHLRTGIRPSTITSSFPQLMSNVDAAYEVAERGTAYFFKGPHYWITRGFQMQGPPRTIYDFGFPRHVQQIDAAVYLREPQKTLFFVGDEYYSYDERKRKMEKDYPKNTEEEFSGVNGQIDAAVELNGYIYFFSGPKTYKYDTEKEDVVSVVKSSSWIGC

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service