Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB1409785

Sigma-Aldrich

Monoclonal Anti-UBTF antibody produced in mouse

clone 6C6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

NOR-90, UBF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C6, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.74 kDa

species reactivity

human, rat

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBTF(7343)

General description

Upstream binding factor (UBF) is a transcription factor required for expression of the 18S, 5.8S, and 28S ribosomal RNAs, along with SL1 (a complex of TBP (MIM 600075) and multiple TBP-associated factors or ′TAFs′). Two UBF polypeptides, of 94 and 97 kD, exist in the human (Bell et al., 1988 [PubMed 3413483]). UBF is a nucleolar phosphoprotein with both DNA binding and transactivation domains. Sequence-specific DNA binding to the core and upstream control elements of the human rRNA promoter is mediated through several HMG boxes (Jantzen et al., 1990 [PubMed 2330041]).[supplied by OMIM

Immunogen

UBTF (NP_055048, 551 a.a. ~ 650 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PPAATNSSKKMKFQGEPKKPPMNGYQKFSQELLSNGELNHLPLKERMVEIGSRWQRISQSQKEHYKKLAEEQQKQYKVHLDLWVKSLSPQDRAAYKEYIS

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service