Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1409494

Sigma-Aldrich

Monoclonal Anti-GIMAP5 antibody produced in mouse

clone 3F11, ascites fluid, solution

Synonym(s):

FLJ11296, HIMAP3, IAN4, IAN4L1, IAN5, IMAP3, hIAN5

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
200 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3F11, monoclonal

form

solution

mol wt

antigen 59.51 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GIMAP5(55340)

General description

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1. (provided by RefSeq)

Immunogen

GIMAP5 (AAH11732, 1 a.a. ~ 307 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGGFQRGKYGTMAEGRSEDNLSATPPALRIILVGKTGCGKSATGNSILGQPAFESKLRAQSVTRTCQVKTGTWNGRKVLVVDTPSIFESQADTQELYKNIGDCYLLSAPGPHVLLLVIQLGRFTAQDTVAIRKVKEVFGTGAMRHVVILFTHKEDLGGQALDDYVANTDNCSLKDLVRECERRYCAFNNWGSVEEQRQQQAELLAVIERLGREREGSFHSNDLFLDAQLLQRTGAGACQEDYRQYQAKVEWQVEKHKQELRENESNWAYKALLRVKHLMLLHYEIFVFLLLCSILFFIIFLFIFHYI

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service