Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1409131

Sigma-Aldrich

Anti-GOLGA1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC33154, golgin-97

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 88.2 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOLGA1(2800)

General description

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren′s syndrome. (provided by RefSeq)

Immunogen

GOLGA1 (AAH32853.1, 1 a.a. ~ 767 a.a) full-length human protein.

Sequence
MFAKLKKKIAEETAVAQRPGGATRIPRSVSKESVASMGADSGDDFASDGSSSREDLSSQLLRRNEQIRKLEARLSDYAEQVRNLQKIKEKLEIALEKHQDSSMRKFQEQNETFQANRAKMAEGLALALARKDQEWSEKMDQLEKEKNILTAQLQEMKNQSMNLFQRRDEMDELEGFQQQELSKIKHMLLKKEESLGKMEQELEARTRELSRTQEELMNSNQMSSDLSQKLEELQRHYSTLEEQRDHVIASKTGAESKITALEQKEQELQALIQQLSIDLQKVTAETQEKEDVITHLQEKVASLEKRLEQNLSGEEHVQELLKEKTLAEQNLEDTRQQLLAARSSQAKAINTLETRVRELEQTLQASEEQLQQSKGIVAAQETQIQELAAANQESSHVQQQALALEQQFLERTQALEAQIVALERMRAADQTTAEQGMRQLEQENAALKECRNEYERSLQNHQFELKKLKEEWSQREIVSVAMAQALEEVRKQREEFQQQAANLTAIIDEKEQNLREKTEVLLQKEQEILQLERGHNSALLQIHQLQAELEALRTLKAEEAAVVAEQEDLLRLRGPLQAEALSVNESHVTSRAMQDPVFQLPTAGRTPNGEVGAMDLTQLQKEKQDLEQQLLEKNKTIKQMQQRMLELRKTLQKELKIRPDNELFEVREKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matteo Marchesini et al.
Cell chemical biology, 27(6), 678-697 (2020-05-11)
The identification of SERCA (sarco/endoplasmic reticulum calcium ATPase) as a target for modulating gain-of-function NOTCH1 mutations in Notch-dependent cancers has spurred the development of this compound class for cancer therapeutics. Despite the innate toxicity challenge associated with SERCA inhibition, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service