Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408665

Sigma-Aldrich

Anti-DUSP14 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MKP-L, MKP6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~22.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DUSP14(11072)

General description

In addition to antigen recognition by the T-cell receptor, T-cell activation requires a second signal from a costimulatory receptor, such as CD28 (MIM 186760), which interacts with B7-1 (CD80; MIM 112203) and B7-2 (CD86; MIM 601020) ligands on antigen-presenting cells. CD28 costimulation induces transcription of interleukin-2 (IL2; MIM 147680) and stabilizes newly synthesized IL2 through the activation of mitogen-activated protein kinases (MAPKs), such as ERK (e.g., MAP2K4; MIM 601335) and JNK (see MIM 601158), and the subsequent creation of AP1 transcription factor (see MIM 165160). DUSP14 is a negative regulator of CD28 signaling.[supplied by OMIM

Immunogen

DUSP14 (NP_008957.1, 1 a.a. ~ 198 a.a) full-length human protein.

Sequence
MSSRGHSTLPRTLMAPRMISEGDIGGIAQITSSLFLGRGSVASNRHLLQARGITCIVNATIEIPNFNWPQFEYVKVPLADMPHAPIGLYFDTVADKIHSVSRKHGATLVHCAAGVSRSATLCIAYLMKFHNVCLLEAYNWVKARRPVIRPNVGFWRQLIDYERQLFGKSTVKMVQTPYGIVPDVYEKESRHLMPYWGI

Application

Anti-DUSP14 antibody produced in rabbit is suitable for western blot analysis.

Biochem/physiol Actions

DUSP14 (Dual specificity phosphatase 14) is associated with several critical signaling pathways. It has ability to dephosphorylate both phosphotyrosine and phosphoserine/phosphothreonine residues on substrates. DUSP14 controls MAPK (mitogen-activated protein kinase) signaling pathway by dephosphorylating MAPK proteins ERK (extracellular-signal-regulated kinase), JNK (c-Jun N-terminal kinase) and p38. It has been reported that DUSP14 participates in T cell proliferation as a negative-feedback regulator.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of Dual-specificity Phosphatase 14 (DUSP14) by PTP Inhibitor V
Bull. Korean Chem. Soc., 34(12), 3871-3873 (2013)
Kate I Patterson et al.
The Biochemical journal, 418(3), 475-489 (2009-02-21)
DUSPs (dual-specificity phosphatases) are a heterogeneous group of protein phosphatases that can dephosphorylate both phosphotyrosine and phosphoserine/phosphothreonine residues within the one substrate. DUSPs have been implicated as major modulators of critical signalling pathways that are dysregulated in various diseases. DUSPs
Y Nakano
The British journal of dermatology, 156(5), 848-860 (2007-02-01)
Nonspecific unresponsive states of delayed-type hypersensitivity (DTH) to unrelated antigens are induced in mice by a single administration of hapten. In these studies, we found a unique regulatory mechanism of contact hypersensitivity (CHS) mediated by nonspecific suppressor factor (NSF) induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service