Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408524

Sigma-Aldrich

Anti-OR3A4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

OLFRA05, OLFRA06, OR17-13, OR17-16, OR17-24, OR17-25, OR24, OR25, OR3A4P

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OR3A4(390756)

Related Categories

General description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. This olfactory receptor gene is transcribed and contains an intact ORF, but it is predicted to be a pseudogene due to a poorly conserved 7-transmembrane domain structure. (provided by RefSeq)

Immunogen

OR3A4 (NP_001005334.1, 1 a.a. ~ 348 a.a) full-length human protein.

Sequence
MDLGNSGNDSVVTKFVLLGLTETAALQPILFVIFLLAYVTTIGGTLSILAAILMETKLHSPMYFFLGNLSLPDVGCVSVTVPAMLSHFISNDRSIPYKACLSELFFFHLLAGADCFLLTIMAYDRYLAICQSLTYSSRMSWGIQQALVGMSCVFSFTNALTQTVALSPLNFCGPNVINHFYCDLPQPFQLSCSSVHLNGQLLFVAAAFMGVAPLVLITVSYAHVAAAVLRIRSAEGRKKAFSTCSSHLTVVGIFYGTGVFSYTRLGSVESSDKDKGIGILNTVISPMLNPLIYWTSLLDVGCISHCSSDAGVSPGPPVQSSLCCLQFTALLSPPPGWGGLSPLNSHGL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark P Dilworth et al.
Annals of surgery, 269(3), 479-485 (2018-02-01)
To investigate differences in methylation between patients with nondysplastic Barrett esophagus who progress to invasive adenocarcinoma and those who do not. Identifying patients with nondysplastic Barrett esophagus who progress to invasive adenocarcinoma remains a challenge. Previous studies have demonstrated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service