Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408301

Sigma-Aldrich

Anti-TBC1D20 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

C20orf140, FLJ45119

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

TBC1D20 (NP_653229.1, 1 a.a. ~ 403 a.a) full-length human protein.

Sequence
MALRSAQGDGPTSGHWDGGAEKADFNAKRKKKVAEIHQALNSDPTDVAALRRMAISEGGLLTDEIRRKVWPKLLNVNANDPPPISGKNLRQMSKDYQQVLLDVRRSLRRFPPGMPEEQREGLQEELIDIILLILERNPQLHYYQGYHDIVVTFLLVVGERLATSLVEKLSTHHLRDFMDPTMDNTKHILNYLMPIIDQVNPELHDFMQSAEVGTIFALSWLITWFGHVLSDFRHVVRLYDFFLAYHPLMPIYFAAVIVLYREQEVLDCDCDMASVHHLLSQIPQDLPYETLISRAGDLFVQFPPSELAREAAAQQQAERTAASTFKDFELASAQQRPDMVLRQRFRGLLRPEDRTKDVLTKPRTNRFVKLAVMGLTVALGAAALAVVKSALEWAPKFQLQLFP

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service