Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408147

Sigma-Aldrich

Anti-ZCCHC3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

C20orf99, MGC104290

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1184


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1184

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZCCHC3(85364)

Related Categories

Immunogen

ZCCHC3 (NP_149080, 1 a.a. ~ 403 a.a) full-length human protein.

Sequence
MATGGGAEEERKRGRPQLLPPARPAARGEEADGGREKMGWAQVVKNLAEKKGEFREPRPPRREEESGGGGGSAGLGGPAGLAAPDLGDFPPAGRGDPKGRRRDPAGEAVDPRKKKGAAEAGRRKKAEAAAAAMATPARPGEAEDAAERPLQDEPAAAAGPGKGRFLVRICFQGDEGACPTRDFVVGALILRSIGMDPSDIYAVIQIPGSREFDVSFRSAEKLALFLRVYEEKREQEDCWENFVVLGRSKSSLKTLFILFRNETVDVEDIVTWLKRHCDVLAVPVKVTDRFGIWTGEYKCEIELRQGEGGVRHLPGAFFLGAERGYSWYKGQPKTCFKCGSRTHMSGSCTQDRCFRCGEEGHLSPYCRKGIVCNLCGKRGHAFAQCPKAVHNSVAAQLTGVAGH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang
Journal of bone oncology, 26, 100335-100335 (2020-11-19)
Osteosarcoma (OS) is a malignant carcinoma often occurring in adolescents. The critical function of long non-coding RNAs (lncRNAs) in cancer arouses increasing attention. Nevertheless, the specific function of FOXD3 Antisense RNA 1 (FOXD3-AS1) in OS has not been understood yet.
Binbin Yi et al.
iScience, 27(3), 109107-109107 (2024-02-22)
Most mammalian cells prevent viral infection and proliferation by expressing various restriction factors and sensors that activate the immune system. Several host restriction factors that inhibit human immunodeficiency virus type 1 (HIV-1) have been identified, but most of them are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service