Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1408001

Sigma-Aldrich

Anti-TRMT2B antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CXorf34, FLJ12687, dJ341D10.3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~56.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRMT2B(79979)

Immunogen

TRMT2B (NP_079193.2, 1 a.a. ~ 504 a.a) full-length human protein.

Sequence
MAGLKRRVPLHSLRYFISMVGLFSKPGLLPWYARNPPGWSQLFLGTVCKGDFTRVIATKCQKGQKSQKKPSHLGPLDGSWQERLADVVTPLWRLSYEEQLKVKFAAQKKILQRLESYIQMLNGVSVTTAVPKSERLSCLLHPIIPSPVINGYRNKSTFSVNRGPDGNPKTVGFYLGTWRDGNVVCVQSNHLKNIPEKHSQVAQYYEVFLRQSPLEPCLVFHEGGYWRELTVRTNSQGHTMAIITFHPQKLSQEELHVQKEIVKEFFIRGPGAACGLTSLYFQESTMTRCSHQQSPYQLLFGEPYIFEELLSLKIRISPDAFFQINTAGAEMLYRTVGELTGVNSDTILLDICCGTGVIGLSLAQHTSRVLGIELLEQAVEDARWTAAFNGITNSEFHTGQAEKILPGLLKSKEDGQSIVAVVNPARAGLHYKVIQAIRNFRAIHTLVFVSCKLHGESTRNVIELCCPPDPAKKLLGEPFVLQQAVPVDLFPHTPHCELVLLFTR

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service