Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1407890

Sigma-Aldrich

Anti-GORASP1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ23443, GOLPH5, GRASP65, MGC118894, MGC118897, P65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~46.5 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GORASP1(64689)

General description

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a membrane protein involved in establishing the stacked structure of the Golgi apparatus. It is a caspase-3 substrate, and cleavage of this encoded protein contributes to Golgi fragmentation in apoptosis. This encoded protein can form a complex with the Golgi matrix protein GOLGA2, and this complex binds to the vesicle docking protein p115. Several alternatively spliced transcript variants of this gene have been identified, but their full-length natures have not been determined. (provided by RefSeq)

Immunogen

GORASP1 (NP_114105.1, 1 a.a. ~ 440 a.a) full-length human protein.

Sequence
MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service