Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1407607

Sigma-Aldrich

Anti-ATG16L1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

APG16L, ATG16L, FLJ00045, FLJ10035, FLJ10828, FLJ22677, IBD10, WDR30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~58.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATG16L1(55054)

General description

Autophagy is the major intracellular degradation system delivering cytoplasmic components to lysosomes, and it accounts for degradation of most long-lived proteins and some organelles. Cytoplasmic constituents, including organelles, are sequestered into double-membraned autophagosomes, which subsequently fuse with lysosomes. ATG16L1 is a component of a large protein complex essential for autophagy (Mizushima et al., 2003 [PubMed 12665549]).[supplied by OMIM

Immunogen

ATG16L1 (NP_110430.4, 1 a.a. ~ 523 a.a) full-length human protein.

Sequence
MAQLRIKHQEELTELHKKRGELAQLVIDLNNQMQRKDREMQMNEAKIAECLQTISDLETECLDLRTKLCDLERANQTLKDEYDALQITFTALEGKLRKTTEENQELVTRWMAEKAQEANRLNAENEKDSRRRQARLQKELAEAAKEPLPVEQDDDIEVIVDETSDHTEETSPVRAISRAATKRLSQPAGGLLDSITNIFGRRSVSSFPVPQDNVDTHPGSGKEVRVPATALCVFDAHDGEVNAVQFSPGSRLLATGGMDRRVKLWEVFGEKCEFKGSLSGSNAGITSIEFDSAGSYLLAASNDFASRIWTVDDYRLRHTLTGHSGKVLSAKFLLDNARIVSGSHDRTLKLWDLRSKVCIKTVFAGSSCNDIVCTEQCVMSGHFDKKIRFWDIRSESIVREMELLGKITALDLNPERTELLSCSRDDLLKVIDLRTNAIKQTFSAPGFKCGSDWTRVVFSPDGSYVAAGSAEGSLYIWSVLTGKVEKVLSKQHSSSINAVAWSPSGSHVVSVDKGCKAVLWAQY

Application

Anti-ATG16L1 antibody produced in mouse is suitable for western blot.

Biochem/physiol Actions

ATG16L1 (autophagy related 16-like 1) is an autophagy-related (ATG) protein. It plays an essential role as a component of the ATG12-ATG5-ATG16L1 complex during autophagy. It functions as a molecular scaffold to form the autophagosome in response to classical and pathogen-related autophagy stimuli. The complex introduces LC3 (ATG8 in yeast) to the autophagosome and connects it to the phosphatidylethanolamine (PE). This conjugation generates a membrane-bound activated form of LC3 named LC3-II.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilio Boada-Romero et al.
The EMBO journal, 32(4), 566-582 (2013-02-05)
Selective autophagy underlies many of the important physiological roles that autophagy plays in multicellular organisms, but the mechanisms involved in cargo selection are poorly understood. Here we describe a molecular mechanism that can target conventional endosomes for autophagic degradation. We
Rhiannon Parkhouse et al.
PloS one, 8(9), e76237-e76237 (2013-10-03)
Autophagy is a fundamental cellular process required for organelle degradation and removal of invasive pathogens. Autophagosome formation involves the recruitment of, and interaction between, multiple proteins produced from autophagy-related (ATG) genes. One of the key complexes in autophagosome formation is
Joëlle Magné et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(5), 1226-1235 (2015-03-15)
Autophagy has emerged as a cell survival mechanism critical for cellular homeostasis, which may play a protective role in atherosclerosis. ATG16L1, a protein essential for early stages of autophagy, has been implicated in the pathogenesis of Crohn's disease. However, it
Vasundhara Kandachar et al.
Journal of cell science, 131(24) (2018-11-09)
The Arf4-rhodopsin complex (mediated by the VxPx motif in rhodopsin) initiates expansion of vertebrate rod photoreceptor cilia-derived light-sensing organelles through stepwise assembly of a conserved trafficking network. Here, we examine its role in the sorting of VAMP7 (also known as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service