Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407360

Sigma-Aldrich

Anti-KCNMB4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNMB4(27345)

General description

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit. (provided by RefSeq)
Potassium calcium activated channel subfamily M regulatory β subunit 4 (KCNMB4) is mapped to human chromosome 12q14.1-15. The encoded protein is a member of the potassium channel β subunit family. KCNMB4 contains two transmembrane segments (TM1 and TM2), intracellular N and C terminal and a large extracellular loop. It is mainly expressed in the brain, neuronal tissue, kidney and bladder smooth muscle.

Immunogen

KCNMB4 (NP_055320.4, 1 a.a. ~ 210 a.a) full-length human protein.

Sequence
MAKLRVAYEYTEAEDKSIRLGLFLIISGVVSLFIFGFCWLSPALQDLQATEANCTVLSVQQIGEVFECTFTCGADCRGTSQYPCVQVYVNNSESNSRALLHSDEHQLLTNPKCSYIPPCKRENQKNLESVMNWQQYWKDEIGSQPFTCYFNQHQRPDDVLLHRTHDEIVLLHCFLWPLVTFVVGVLIVVLTICAKSLAVKAEAMKKRKFS

Biochem/physiol Actions

Potassium calcium activated channel subfamily M regulatory α subunit 4 (KCNMB4) plays a vital role in increasing Ca2+ sensitivity. It reduces the voltage dependence of the conductance-voltage relationship as well as the slope of the gating charge-voltage curve.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological consequences of the coexpression of BK channel α and auxiliary β subunits
Torres YP, et al.
Frontiers in Physiology, 5, 383-383 (2014)
hKCNMB3 and hKCNMB4, cloning and characterization of two members of the large?conductance calcium?activated potassium channel β subunit family
Behrens R, et al.
Febs Letters, 474(1), 99-106 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service