Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1407350

Sigma-Aldrich

Anti-KLHL20 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

KHLHX, KLEIP, KLHLX, RP3-383J4.3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~68 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLHL20(27252)

General description

The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 44-56 amino acid repeat motif. The kelch motif appears in many different polypeptide contexts and contains multiple potential protein-protein contact sites. Members of this family are present both throughout the cell and extracellularly, with diverse activities. (provided by RefSeq)

Immunogen

KLHL20 (NP_055273.2, 1 a.a. ~ 609 a.a) full-length human protein.

Sequence
MEGKPMRRCTNIRPGETGMDVTSRCTLGDPNKLPEGVPQPARMPYISDKHPRQTLEVINLLRKHRELCDVVLVVGAKKIYAHRVILSACSPYFRAMFTGELAESRQTEVVIRDIDERAMELLIDFAYTSQITVEEGNVQTLLPAACLLQLAEIQEACCEFLKRQLDPSNCLGIRAFADTHSCRELLRIADKFTQHNFQEVMESEEFMLLPANQLIDIISSDELNVRSEEQVFNAVMAWVKYSIQERRPQLPQVLQHVRLPLLSPKFLVGTVGSDPLIKSDEECRDLVDEAKNYLLLPQERPLMQGPRTRPRKPIRCGEVLFAVGGWCSGDAISSVERYDPQTNEWRMVASMSKRRCGVGVSVLDDLLYAVGGHDGSSYLNSVERYDPKTNQWSSDVAPTSTCRTSVGVAVLGGFLYAVGGQDGVSCLNIVERYDPKENKWTRVASMSTRRLGVAVAVLGGFLYAVGGSDGTSPLNTVERYNPQENRWHTIAPMGTRRKHLGCAVYQDMIYAVGGRDDTTELSSAERYNPRTNQWSPVVAMTSRRSGVGLAVVNGQLMAVGGFDGTTYLKTIEVFDPDANTWRLYGGMNYRRLGGGVGVIKMTHCESHIW

Biochem/physiol Actions

KLHL20 (kelch like family member 20) is a substrate adaptor of cullin3 (Cul3). It is required for post-Golgi trafficking. The Cul3-KLHL20 complex works as an ubiquitin E3 ligase and is responsible for polyubiquitination of coronin-7, thereby targeting coronin-7 to trans-Golgi network. KLHL20 is also involved in autophagy by regulating autophagy-associated degradation of ULK1 (unc-51 like autophagy activating kinase 1) and VPS34 (vacuolar protein sorting 34) complex subunits. Additionally, KLHL20 is associated with hypoxia responses and tumorigenesis. HIF1 (hypoxia inducible factor 1)-mediated upregulation of KLHL20 gene causes hypoxia-induced PML (promyelocytic leukemia) polyubiquitination. PML destruction is linked with epithelial-mesenchymal transition, tumor growth, angiogenesis and other related events.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Chien Yuan et al.
Molecular cell, 54(4), 586-600 (2014-04-29)
Ubiquitin chains are formed as structurally distinct polymers via different linkages, and several chain types including K33-linkage remain uncharacterized. Here, we describe a role for K33-polyubiquitination in protein trafficking. We show that the Cullin 3 (Cul3) substrate adaptor KLHL20 is
Chin-Chih Liu et al.
Molecular cell, 61(1), 84-97 (2015-12-22)
Autophagy, a cellular self-eating mechanism, is important for maintaining cell survival and tissue homeostasis in various stressed conditions. Although the molecular mechanism of autophagy induction has been well studied, how cells terminate autophagy process remains elusive. Here, we show that
Wei-Chien Yuan et al.
Cancer cell, 20(2), 214-228 (2011-08-16)
Tumor hypoxia is associated with disease progression and treatment failure, but the hypoxia signaling mechanism is not fully understood. Here, we show that KLHL20, a Cullin3 (Cul3) substrate adaptor induced by HIF-1, coordinates with the actions of CDK1/2 and Pin1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service