SAB1407228
Anti-ZNF346 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Synonym(s):
DKFZp547M223, JAZ, Zfp346
Select a Size
$541.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$541.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~32.9 kDa
species reactivity
human
technique(s)
indirect immunofluorescence: suitable
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ZNF346(23567)
General description
Immunogen
Sequence
MEYPAPATVQAADGGAAGPYSSSELLEGQEPDGVRFDRERARRLWEAVSGAQPVGREEVEHMIQKNQCLFTNTQCKVCCALLISESQKLAHYQSKKHANKVKRYLAIHGMETLKGETKKLDSDQKSSRSKDKNQCCPICNMTFSSPVVAQSHYLGKTHAKNLKLKQQSTKVEALHQNREMIDPDKFCSLCHATFNDPVMAQQHYVGKKHRKQETKLKLMARYGRLADPAVTDFPAGKGYPCKTCKIVLNSIEQYQAHVSGFKHKNQSPKTVASSLGQIPMQRQPIQKDSTTLED
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service