Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1406529

Sigma-Aldrich

Anti-THBS4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

TSP4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~105.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... THBS4(7060)

General description

Thrombospondin 4 (THBS4) gene is located on human chromosome location 5q14.1.THBS4 is a secreted multidomain glycoprotein of the extracellular matrix.

Immunogen

THBS4 (NP_003239.2, 1 a.a. ~ 961 a.a) full-length human protein.

Sequence
MLAPRGAAVLLLHLVLQRWLAAGAQATPQVFDLLPSSSQRLNPGALLPVLTDPALNDLYVISTFKLQTKSSATIFGLYSSTDNSKYFEFTVMGRLNKAILRYLKNDGKVHLVVFNNLQLADGRRHRILLRLSNLQRGAGSLELYLDCIQVDSVHNLPRAFAGPSQKPETIELRTFQRKPQDFLEELKLVVRGSLFQVASLQDCFLQQSEPLAATGTGDFNRQFLGQMTQLNQLLGEVKDLLRQQVKETSFLRNTIAECQACGPLKFQSPTPSTVVPPAPPAPPTRPPRRCDSNPCFRGVQCTDSRDGFQCGPCPEGYTGNGITCIDVDECKYHPCYPGVHCINLSPGFRCDACPVGFTGPMVQGVGISFAKSNKQVCTDIDECRNGACVPNSICVNTLGSYRCGPCKPGYTGDQIRGCKAERNCRNPELNPCSVNAQCIEERQGDVTCVCGVGWAGDGYICGKDVDIDSYPDEELPCSARNCKKDNCKYVPNSGQEDADRDGIGDACDEDADGDGILNEQDNCVLIHNVDQRNSDKDIFGDACDNCLSVLNNDQKDTDGDGRGDACDDDMDGDGIKNILDNCPKFPNRDQRDKDGDGVGDACDSCPDVSNPNQSDVDNDLVGDSCDTNQDSDGDGHQDSTDNCPTVINSAQLDTDKDGIGDECDDDDDNDGIPDLVPPGPDNCRLVPNPAQEDSNSDGVGDICESDFDQDQVIDRIDVCPENAEVTLTDFRAYQTVVLDPEGDAQIDPNWVVLNQGMEIVQTMNSDPGLAVGYTAFNGVDFEGTFHVNTQTDDDYAGFIFGYQDSSSFYVVMWKQTEQTYWQATPFRAVAEPGIQLKAVKSKTGPGEHLRNSLWHTGDTSDQVRLLWKDSRNVGWKDKVSYRWFLQHRPQVGYIRVRFYEGSELVADSGVTIDTTMRGGRLGVFCFSQENIIWSNLKYRCNDTIPEDFQEFQTQNFDRFDN

Biochem/physiol Actions

Thrombospondin 4 (THBS4) plays an important role in cellular migration and proliferation.THBS4 acts as a tumor suppressor gene. THBS4 methylation is considered as a biomarker for the occurrence of adenomas and carcinoma.THBS4 expression in breast cancer-associated extracellular matrix facilitates the invasion of tumor cells.THBS4 is used as a powerful marker for diffuse-type gastric adenocarcinomas.Inhibition of THBS4 inhibits prostate cancer (PCa) cell migration and invasion.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gastric precancerous lesions are associated with gene variants in Helicobacter pylori-susceptible ethnic Malays
Maran S, et al.
World Journal of Gastroenterology, 19(23), 3615-3615 (2013)
Thrombospondin-4 expression is activated during the stromal response to invasive breast cancer
Reed A, et al.
Virchows Archiv, 463(4), 535-545 (2013)
Reciprocal regulation of long noncoding RNAs THBS4-003 and THBS4 control migration and invasion in prostate cancer cell lines
Liu J, et al.
Molecular Medicine Reports, 14(2), 1451-1458 (2016)
Thrombospondin-4 is a putative tumour-suppressor gene in colorectal cancer that exhibits age-related methylation
Greco S, et al.
BMC Cancer, 10(1), 494-494 (2010)
THBS4, a novel stromal molecule of diffuse-type gastric adenocarcinomas, identified by transcriptome-wide expression profiling
Forster S, et al.
Modern Pathology, 24(10), 1390-1390 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service