Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1406394

Sigma-Aldrich

Anti-RNF6 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686P0776

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~78.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF6(6049)

General description

The protein encoded by this gene contains a RING-H2 finger motif. Deletions and mutations in this gene were detected in esophageal squamous cell carcinoma (ESCC), suggesting that this protein may be a potential tumor suppressor. Studies of the mouse counterpart suggested a role of this protein in the transcription regulation that controls germinal differentiation. Multiple alternatively spliced transcript variants encoding the same protein are observed. (provided by RefSeq)

Immunogen

RNF6 (NP_005968.1, 1 a.a. ~ 685 a.a) full-length human protein.

Sequence
MNQSRSRSDGGSEETLPQDHNHHENERRWQQERLHREEAYYQFINELNDEDYRLMRDHNLLGTPGEITSEELQQRLDGVKEQLASQPDLRDGTNYRDSEVPRESSHEDSLLEWLNTFRRTGNATRSGQNGNQTWRAVSRTNPNNGEFRFSLEIHVNHENRGFEIHGEDYTDIPLSDSNRDHTANRQQRSTSPVARRTRSQTSVNFNGSSSNIPRTRLASRGQNPAEGSFSTLGRLRNGIGGAAGIPRANASRTNFSSHTNQSGGSELRQREGQRFGAAHVWENGARSNVTVRNTNQRLEPIRLRSTSNSRSRSPIQRQSGTVYHNSQRESRPVQQTTRRSVRRRGRTRVFLEQDRERERRGTAYTPFSNSRLVSRITVEEGEESSRSSTAVRRHPTITLDLQVRRIRPGENRDRDSIANRTRSRVGLAENTVTIESNSGGFRRTISRLERSGIRTYVSTITVPLRRISENELVEPSSVALRSILRQIMTGFGELSSLMEADSESELQRNGQHLPDMHSELSNLGTDNNRSQHREGSSQDRQAQGDSTEMHGENETTQPHTRNSDSRGGRQLRNPNNLVETGTLPILRLAHFFLLNESDDDDRIRGLTKEQIDNLSTRHYEHNSIDSELGKICSVCISDYVTGNKLRQLPCMHEFHIHCIDRWLSENCTCPICRQPVLGSNIANNG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service