Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1406221

Sigma-Aldrich

Anti-PAX6 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AN, AN2, D11S812E, MGC17209, MGDA, WAGR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~46.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAX6(5080)

General description

The paired box 6 (PAX6) gene with 14 exons, is mapped to human chromosome 11p13. The encoded protein belongs to the paired-domain Pax family. PAX6 is a 422 amino acids protein, generally expressed in the developing eye, both in neuroectoderm and surface ectoderm. It is also present in the adult eye.

Immunogen

PAX6 (NP_000271.1, 1 a.a. ~ 422 a.a) full-length human protein.

Sequence
MQNSHSGVNQLGGVFVNGRPLPDSTRQKIVELAHSGARPCDISRILQVSNGCVSKILGRYYETGSIRPRAIGGSKPRVATPEVVSKIAQYKRECPSIFAWEIRDRLLSEGVCTNDNIPSVSSINRVLRNLASEKQQMGADGMYDKLRMLNGQTGSWGTRPGWYPGTSVPGQPTQDGCQQQEGGGENTNSISSNGEDSDEAQMRLQLKRKLQRNRTSFTQEQIEALEKEFERTHYPDVFARERLAAKIDLPEARIQVWFSNRRAKWRREEKLRNQRRQASNTPSHIPISSSFSTSVYQPIPQPTTPVSSFTSGSMLGRTDTALTNTYSALPPMPSFTMANNLPMQPPVPSQTSSYSCMLPTSPSVNGRSYDTYTPPHMQTHMNSQPMGTSGTTSTGLISPGVSVPVQVPGSEPDMSQYWPRLQ

Biochem/physiol Actions

Paired box 6 (PAX6) is a highly conserved transcriptional regulator involved in oculogenesis. In addition, it is also implicated in maintenance of adult eye. Mutation in the gene leads to the development of aniridia in humans and small eyes in rats.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic structure, evolutionary conservation and aniridia mutations in the human PAX6 gene.
Glaser T, et al.
Nature Genetics, 2(3), 232-232 (1992)
Experimental assessment of novel PAX6 splicing mutations in two Chinese families with aniridia.
Miao Q, et al.
Gene, 630, 44-48 (2017)
A susceptibility locus for myopia in the normal population is linked to the PAX6 gene region on chromosome 11: a genomewide scan of dizygotic twins.
Hammond C J, et al.
American Journal of Human Genetics, 75(2), 294-304 (2004)
Newly identified paired box 6 mutation of variant familial aniridia: Congenital iris ectropion with foveal hypoplasia.
Kim W J, et al.
Indian Journal of Ophthalmology, 65(1), 55-55 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service