Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1406138

Sigma-Aldrich

Anti-MOG antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC26137, MOGIG-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~32.56 kDa

species reactivity

rat, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MOG(4340)

General description

The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintenance of the myelin sheath and in cell-cell communication. Alternatively spliced transcript variants encoding different isoforms have been identified. (provided by RefSeq)

Immunogen

MOG (AAH35938, 1 a.a. ~ 295 a.a) full-length human protein.

Sequence
MASLSRPSLPSCLCSFLLLLLLQVSSSYAGQFRVIGPRHPIRALVGDEVELPCRISPGKNATGMEVGWYRPPFSRVVHLYRNGKDQDGDQAPEYRGRTELLKDAIGEGKVTLRIRNVRFSDEGGFTCFFRDHSYQEEAAMELKVEDPFYWVSPGVLVLLAVLPVLLLQITVGLVFLCLQYRLRGKLRAEIENLHRTFDPHFLRVPCWKITLFVIVPVLGPLVALIICYNWLHRRLAGQFLEELRKFSSLCYKQRIKSQERETEATRGRGGLLRDHIPRGKEELESLGGGKTPPGR

Application

Anti-MOG antibody produced in mouse is suitable for western blot analysis.

Biochem/physiol Actions

Myelin/oligodendrocyte glycoprotein (MOG) is a major component of the central nervous system. It is involved in several cellular processes including cellular adhesion, in the regulation of oligodendrocyte microtubule stability and facilitates the interactions between myelin and the immune system. It has been reported that MOG may be involved in the completion and/or maintenance of the myelin sheath, and in cell-cell communication. It has also been observed that mutation or polymorphisms in MOG gene could cause multiple sclerosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth B Hutchinson et al.
Shock (Augusta, Ga.), 46(3 Suppl 1), 167-176 (2016-06-14)
During the acute time period following traumatic brain injury (TBI), noninvasive brain imaging tools such as magnetic resonance imaging (MRI) can provide important information about the clinical and pathological features of the injury and may help predict long-term outcomes. In
Dan Benjamini et al.
NeuroImage, 221, 117195-117195 (2020-07-30)
We describe a practical two-dimensional (2D) diffusion MRI framework to deliver specificity and improve sensitivity to axonal injury in the spinal cord. This approach provides intravoxel distributions of correlations of water mobilities in orthogonal directions, revealing sub-voxel diffusion components. Here
Dan Benjamini et al.
NeuroImage, 163, 183-196 (2017-09-26)
Magnetic resonance imaging (MRI) provides a powerful set of tools with which to investigate biological tissues noninvasively and in vivo. Tissues are heterogeneous in nature; an imaging voxel contains an ensemble of different cells and extracellular matrix components. A long-standing
B della Gaspera et al.
European journal of biochemistry, 258(2), 478-484 (1999-01-05)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the mammalian central nervous system, is located on the surface of the oligodendrocyte plasma membrane and the outermost lamellae of mature myelin; it is expressed during the latter steps of myelinogenesis. It has
D Pham-Dinh et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 7990-7994 (1993-09-01)
Myelin/oligodendrocyte glycoprotein (MOG) is found on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system, and it is a target antigen in experimental autoimmune encephalomyelitis and multiple sclerosis. We have isolated bovine, mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service