Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1405800

Sigma-Aldrich

Anti-Fas Antibody

mouse polyclonal

Synonym(s):

CD64, CD64A, FCRI, FLJ18345, IGFR1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

Product Name

Anti-FCGR1A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~42.6 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FCGR1A(2209)

General description

Fc fragment of IgG receptor Ia (FCGR1A) gene encodes a protein that plays an important role in the immune response. This protein is a high-affinity Fc-gamma receptor. The gene is one of three related gene family members located on chromosome 1. (provided by RefSeq). Fc fragment of IgG receptor Ia (FCGR1A) gene is located on human chromosome 1q21.2. FCGR1A is expressed on monocytes and macrophages.

Immunogen

FCGR1A (NP_000557.1, 1 a.a. ~ 374 a.a) full-length human protein.

Sequence
MWFLTTLLLWVPVDGQVDTTKAVITLQPPWVSVFQEETVTLHCEVLHLPGSSSTQWFLNGTATQTSTPSYRITSASVNDSGEYRCQRGLSGRSDPIQLEIHRGWLLLQVSSRVFTEGEPLALRCHAWKDKLVYNVLYYRNGKAFKFFHWNSNLTILKTNISHNGTYHCSGMGKHRYTSAGISVTVKELFPAPVLNASVTSPLLEGNLVTLSCETKLLLQRPGLQLYFSFYMGSKTLRGRNTSSEYQILTARREDSGLYWCEAATEDGNVLKRSPELELQVLGLQLPTPVWFHVLFYLAVGIMFLVNTVLWVTIRKELKRKKKWDLEISLDSGHEKKVISSLQEDRHLEEELKCQEQKEEQLQEGVHRKEPQGAT

Biochem/physiol Actions

Fc fragment of IgG receptor Ia (FCGR1A) functions as the single high-affinity receptor of immunoglobulin G (IgG) in humans. It also regulates immune inflammation. FCGR1A acts as a diagnostic marker for bacterial infections, such as neonatal sepsis. FCGR1A is an important biomarker for rheumatoid arthritis (RA).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of human specific gene duplications relative to other primates by array CGH and quantitative PCR
Armengol G, et al.
Genomics, 95(4) (2010)
Neutrophil CD64 as a diagnostic marker for neonatal sepsis: Meta-analysis.
Dai J, et al.
Advances in Clinical and Experimental Medicine : Official Organ Wroclaw Medical University (2017)
FcγRI (CD64) contributes to the severity of immune inflammation through regulating NF-κB/NLRP3 inflammasome pathway
Zhang H, et al.
Life Sciences (2018)
Elevated membrane and soluble CD64: a novel marker reflecting altered FcγR function and disease in early rheumatoid arthritis that can be regulated by anti-rheumatic treatment
Matt P, et al.
PLoS ONE, 10(9), e0137474-e0137474 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service