Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405756

Sigma-Aldrich

Anti-EMR1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90.4 kDa

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... EMR1(2015)

General description

This gene encodes a protein that has a domain resembling seven transmembrane G protein-coupled hormone receptors (7TM receptors) at its C-terminus. The N-terminus of the encoded protein has six EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. (provided by RefSeq)

Immunogen

EMR1 (AAH59395, 1 a.a. ~ 821 a.a) full-length human protein.

Sequence
MRGFNLLLFWGCCVMHSWEGHIRPTRKPNTKGNNCRDSTLCPAYATCTNTVDSYYCACKQGFLSSNGQNHFKDPGVRCKDIDECSQSPQPCGPNSSCKNLSGRYKCSCLDGFSSPTGNDWVPGKPGNFSCTDINECLTSSVCPEHSDCVNSMGSYSCSCQVGFISRNSTCEDVDECADPRACPEHATCNNTVGNYSCFCNPGFESSSGHLSFQGLKASCEDIDECTEMCPINSTCTNTPGSYFCTCHPGFAPSNGQLNFTDQGVECRDIDECRQDPSTCGPNSICTNALGSYSCGCIAGFHPNPEGSQKDGNFSCQRVLFKCKEDVIPDNKQIQQCQEGTAVKPAYVSFCAQINNIFSVLDKVCENKTTVVSLKNTTESFVPVLKQISTWTKFTKEETSSLATVFLESVESMTLASFWKPSANITPAVRTEYLDIESKVINKECSEENVTLDLVAKGDKMKIGCSTIEESESTETTGVAFVSFVGMESVLNERFFKDHQAPLTTSEIKLKMNSRVVGGIMTGEKKDGFSDPIIYTLENIQPKQKFERPICVSWSTDVKGGRWTSFGCVILEASETYTICSCNQMANLAIIMASGELTMGCAIIAGFLHYLFLACFFWMLVEAVILFLMVRNLKVVNYFSSRNIKMLHICAFGYGLPMLVVVISASVQPQGYGMHNRCWLNTETGFIWSFLGPVCTVIVINSLLLTWTLWILRQRLSSVNAEVSTLKDTRLLTFKAFAQLFILGCSWVLGIFQIGPVAGVMAYLFTIINSLQGAFIFLIHCLLNGQVREEYKRWITGKTKPSSQSQTSRILLSSMPSASKTG

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyo Kashiwagi et al.
Vaccine, 32(27), 3393-3401 (2014-04-29)
Aluminum-adjuvanted vaccines are administered through an intramuscular injection (IM) in the US and EU, however, a subcutaneous injection (SC) has been recommended in Japan because of serious muscle contracture previously reported following multiple IMs of antibiotics. Newly introduced adjuvanted vaccines
Łukasz Bułdak et al.
Experimental and therapeutic medicine, 11(3), 1095-1103 (2016-03-22)
The treatment of diabetes and its complications is a key challenge for healthcare professionals. Accelerated atherosclerosis is associated with progressive diabetes, and it has been indicated that macrophages serve a crucial function in this process. Currently, the first-line treatment of
Flávio V Loures et al.
The Journal of infectious diseases, 210(5), 762-773 (2014-03-08)
Dectin-1, the innate immune receptor that recognizes β-glucan, plays an important role in immunity against fungal pathogens. Paracoccidioides brasiliensis, the etiological agent of paracoccidioidomycosis, has a sugar-rich cell wall mainly composed of mannans and glucans. This fact motivated us to
Naoyuki Kawao et al.
PloS one, 10(4), e0123982-e0123982 (2015-04-22)
Macrophages play crucial roles in repair process of various tissues. However, the details in the role of macrophages during bone repair still remains unknown. Herein, we examined the contribution of the tissue fibrinolytic system to the macrophage functions in bone
Jianzhong Li et al.
Kidney international, 86(1), 86-102 (2014-01-24)
The mammalian target of rapamycin (mTOR) plays a critical role for cell growth and survival in many cell types. While substantial progress has been made in understanding the abnormal activation of mTORC1 in the pathogenesis of kidney disease, little is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service