Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405570

Sigma-Aldrich

Anti-CCNA2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CCN1, CCNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48.5 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNA2(890)

General description

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. In contrast to cyclin A1, which is present only in germ cells, this cyclin is expressed in all tissues tested. This cyclin binds and activates CDC2 or CDK2 kinases, and thus promotes both cell cycle G1/S and G2/M transitions. (provided by RefSeq)

Immunogen

CCNA2 (NP_001228.1, 1 a.a. ~ 432 a.a) full-length human protein.

Sequence
MLGNSAPGPATREAGSALLALQQTALQEDQENINPEKAAPVQQPRTRAALAVLKSGNPRGLAQQQRPKTRRVAPLKDLPVNDEHVTVPPWKANSKQPAFTIHVDEAEKEAQKKPAESQKIEREDALAFNSAISLPGPRKPLVPLDYPMDGSFESPHTMDMSIVLEDEKPVSVNEVPDYHEDIHTYLREMEVKCKPKVGYMKKQPDITNSMRAILVDWLVEVGEEYKLQNETLHLAVNYIDRFLSSMSVLRGKLQLVGTAAMLLASKFEEIYPPEVAEFVYITDDTYTKKQVLRMEHLVLKVLTFDLAAPTVNQFLTQYFLHQQPANCKVESLAMFLGELSLIDADPYLKYLPSVIAGAAFHLALYTVTGQSWPESLIRKTGYTLESLKPCLMDLHQTYLKAPQHAQQSIREKYKNSKYHGVSLLNPPETLNL

Biochem/physiol Actions

Cyclin A2 (CCNA2) is a marker for estrogen receptor positive (ER+) breast cancer. It has a role in cell cycle control at the G1/S and the G2/M transitions. The protein maybe involved in epithelial-mesenchymal transitions (EMT) and metastasis. CCNA2 has been shown to be upregulated in many cancers.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luís Muñoz-Bellvis et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 25(4), 590-601 (2012-01-10)
Most genetic studies in colorectal carcinomas have focused on those abnormalities that are acquired by primary tumors, particularly in the transition from adenoma to carcinoma, whereas few studies have compared the genetic abnormalities of primary versus paired metastatic samples. In
Tian Gao et al.
PloS one, 9(3), e91771-e91771 (2014-03-14)
Identification of effective prognostic biomarkers and targets are of crucial importance to the management of estrogen receptor positive (ER+) breast cancer. CCNA2 (also known as CyclinA2) belongs to the highly conserved cyclin family and is significantly overexpressed in various cancer
Arata Honda et al.
PloS one, 7(6), e39249-e39249 (2012-06-30)
Human cyclin A2 is a key regulator of S phase progression and entry into mitosis. Alternative splice variants of the G1 and mitotic cyclins have been shown to interfere with full-length cyclin functions to modulate cell cycle progression and are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service