Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1405428

Sigma-Aldrich

Anti-ADFP antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADRP, MGC10598

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48.1 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADFP(123)

General description

Adipocyte differentiation-related protein is associated with the globule surface membrane material. This protein is a major constituent of the globule surface. Increase in mRNA levels is one of the earliest indications of adipocyte differentiation (provided by RefSeq)
Adipose differentiation-related protein (ADRP), also known as adipose differentiation-related protein (ADFP), is the major lipid droplet protein. It is encoded by the gene mapped to human chromosome 9p22.1–p21.3. ADFP is ubiquitously expressed and is a member of PAT (perilipin/APRP/TIP47) family.

Immunogen

ADFP (NP_001113.2, 1 a.a. ~ 437 a.a) full-length human protein.

Sequence
MASVAVDPQPSVVTRVVNLPLVSSTYDLMSSAYLSTKDQYPYLKSVCEMAENGVKTITSVAMTSALPIIQKLEPQIAVANTYACKGLDRIEERLPILNQPSTQIVANAKGAVTGAKDAVTTTVTGAKDSVASTITGVMDKTKGAVTGSVEKTKSVVSGSINTVLGSRMMQLVSSGVENALTKSELLVEQYLPLTEEELEKEAKKVEGFDLVQKPSYYVRLGSLSTKLHSRAYQQALSRVKEAKQKSQQTISQLHSTVHLIEFARKNVYSANQKIQDAQDKLYLSWVEWKRSIGYDDTDESHCAEHIESRTLAIARNLTQQLQTTCHTLLSNIQGVPQNIQDQAKHMGVMAGDIYSVFRNAASFKEVSDSLLTSSKGQLQKMKESLDDVMDYLVNNTPLNWLVGPFYPQLTESQNAQDQGAEMDKSSQETQRSEHKTH

Biochem/physiol Actions

Adipose differentiation-related protein (ADFP) plays a vital role in lipid droplet (LD) formation and/or maturation. Increased expression of this protein in tamm-horsfall protein 1 (THP-1) macrophages increases lipid accumulation and stops lipid efflux. In addition, ADFP is slightly involved in maintenance of adipose tissue homeostasis. It acts as a potential marker for intracellular LD in tissues.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of Triglyceride Metabolism. III. Emerging role of lipid droplet protein ADFP in health and disease
Chang BHJ, et al.
The American Journal of Physiology, 292(6), G1465-G1468 (2007)
Functional Compensation for Adipose Differentiation-related Protein (ADFP) by Tip47 in an ADFP Null Embryonic Cell Line
Sztalryd C, et al.
The Journal of Biological Chemistry, 281(45), 34341-34348 (2006)
Identification of differentially expressed genes in omental adipose tissues of obese patients by suppression subtractive hybridization
Qiu J, et al.
Biochemical and Biophysical Research Communications, 352(2), 469-478 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service