Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1404174

Sigma-Aldrich

Monoclonal Anti-PCDHGC3 antibody produced in mouse

clone 1C4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PC43, PCDH-GAMMA-C3, PCDH2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~128.85 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDHGC3(5098)

General description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. (provided by RefSeq)

Immunogen

PCDHGC3 (AAH26218.1, 1 a.a. ~ 934 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVPEAWRSGLVSTGRVVGVLLLLGALNKASTVIHYEIPEEREKGFAVGNVVANLGLDLGSLSARRFRVVSGASRRFFEVNRETGEMFVNDRLDREELCGTLPSCTVTLELVVENPLELFSVEVVIQDINDNNPAFPTQEMKLEISEAVAPGTRFPLESAHDPDVGSNSLQTYELSRNEYFALRVQTREDSTKYAELVLERALDREREPSLQLVLTALDGGTPALSASLPIHIKVLDANDNAPVFNQSLYRARVLEDAPSGTRVVQVLATDLDEGPNGEIIYSFGSHNRAGVRQLFALDLVTGMLTIKGRLDFEDTKLHEIYIQAKDKGANPEGAHCKVLVEVVDVNDNAPEITVTSVYSPVPEDAPLGTVIALLSVTDLDAGENGLVTCEVPPGLPFSLTSSLKNYFTLKTSADLDRETVPEYNLSITARDAGTPSLSALTIVRVQVSDINDNPPQSSQSSYDVYIEENNLPGAPILNLSVWDPDAPQNARLSFFLLEQGAETGLVGRYFTINRDNGIVSSLVPLDYEDRREFELTAHISDGGTPVLATNISVNIFVTDRNDNAPQVLYPRPGGSSVEMLPRGTSAGHLVSRVVGWDADAGHNAWLSYSLLGSPNQSLFAIGLHTGQISTARPVQDTDSPRQTLTVLIKDNGEPSLSTTATLTVSVTEDSPEARAEFPSGSAPREQKKNLTFYLLLSLILVSVGFVVTVFGVIIFKVYKWKQSRDLYRAPVSSLYRTPGPSLHADAVRGGLMSPHLYHQVYLTTDSRRSDPLLKKPGAASPLASRQNTLRSCDPVFYRQVLGAESAPPGQQAPPNTDWRFSQAQRPGTSGSQNGDDTGTWPNNQFDTEMLQAMILASASEAADGSSTLGGGAGTMGLSARYGPQFTLQHVPDYRQNVYIPGSNATLTNAAGKRDGKAPAGGNGNKKKSGKKEKK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service