Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1402502

Sigma-Aldrich

Monoclonal Anti-CHST4 antibody produced in mouse

clone 4D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

LSST

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHST4(10164)

Immunogen

CHST4 (NP_005760, 221 a.a. ~ 320 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LMIDSRIVMGQHEQKLKKEDQPYYVMQVICQSQLEIYKTIQSLPKALQERYLLVRYEDLARAPVAQTSRMYEFVGLEFLPHLQTWVHNITRGKGMGDHAF

Biochem/physiol Actions

CHST4 (Carbohydrate sulfotransferase 4) is a sulfotransferase which plays a vital role in L-selectin ligand formation in the vascular endothelial cells. It possesses GlcNAc-6-sulfotransferase activity. CHST4 transfers sulfate from 3′-phosphoadenosine 5′ phospho-sulfate to the 6-hydroxyl group of N-acetylglucosamine on glycoproteins.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José L Pablos et al.
BMC immunology, 6, 6-6 (2005-03-09)
The recruitment of lymphocytes to secondary lymphoid organs relies on interactions of circulating cells with high endothelial venules (HEV). HEV are exclusive to these organs under physiological conditions, but they can develop in chronically-inflamed tissues. The interaction of L-selectin on
X Li et al.
Journal of leukocyte biology, 69(4), 565-574 (2001-04-20)
Sulfation is an essential component of the selectin ligands, potentially mediated by members of a new family of carbohydrate sulfotransferases. In this study, we assessed the contributions of CHST1, CHST2, CHST3, and CHST4 in producing functional L-selectin ligands. Human umbilical
A Bistrup et al.
The Journal of cell biology, 145(4), 899-910 (1999-05-20)
L-selectin, a lectin-like receptor, mediates rolling of lymphocytes on high endothelial venules (HEVs) in secondary lymphoid organs by interacting with HEV ligands. These ligands consist of a complex of sialomucins, candidates for which are glycosylation- dependent cell adhesion molecule 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service