Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB1402428

Sigma-Aldrich

Monoclonal Anti-USO1 antibody produced in mouse

clone 3F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

P115, TAP, VDP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~131.82 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USO1(8615)

General description

The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner. (provided by RefSeq)

Immunogen

USO1 (AAH32654, 1 a.a. ~ 961 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MNFLRGVMGGQSAGPQHTEAETIQKLCDRVASSTLLDDRRNAVRALKSLSKKYRLEVGIQAMEHLIHVLQTDRSDSEIIGYALDTLYNIISNEEEEVEENSTRQSEDLGSQFTEIFIKQQENVTLLLSLLEEFDFHVRWPGVKLLTSLLKQLGPQVQQIILVSPMGVSRLMDLLADSREVIRNDGVLLLQALTRSNGAIQKIVAFENAFERLLDIISEEGNSDGGIVVEDCLILLQNLLKNNNSNQNFFKEGSYIQRMKPWFEVGDENSGWSAQKVTNLHLMLQLVRVLVSPTNPPGATSSCQKAMFQCGLLQQLCTILMATGVPADILTETINTVSEVIRGCQVNQDYFASVNAPSNPPRPAIVVLLMSMVNERQPFVLRCAVLYCFQCFLYKNQKGQGEIVSTLLPSTIDATGNSVSAGQLLCGGLFSTDSLSNWCAAVALAHALQENATQKEQLLRVQLATSIGNPPVSLLQQCTNILSQGSKIQTRVGLLMLLCTWLSNCPIAVTHFLHNSANVPFLTGQIAENLGEEEQLVQGLCALLLGISIYFNDNSLESYMKEKLKQLIEKRIGKENFIEKLGFISKHELYSRASQKPQPNFPSPEYMIFDHEFTKLVKELEGVITKAIYKSSEEDKKEEEVKKTLEQHDNIVTHYKNMIREQDLQLEELRQQVSTLKCQNEQLQTAVTQQVSQIQQHKDQYNLLKIQLGKDNQHQGSYSEGAQMNGIQPEEIGRLREEIEELKRNQELLQSQLTEKDSMIENMKSSQTSGTNEQSSAIVSARDSEQVAELKQELATLKSQLNSQSVEITKLQTEKQELLQKTEAFAKSVEVQGETETIIATKTTDVEGRLSALLQETKELKNEIKALSEERTAIKEQLDSSNSTIAILQTEKDKLELEITDSKKEQDDLLVLLADQDQKILSLKNKLKDLGHPVEEEDELESGDQEDEDDESEDPGKDLDHI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service