Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1402417

Sigma-Aldrich

Monoclonal Anti-FZD7 antibody produced in mouse

clone 4D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

FzE3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD7(8324)

General description

Frizzled class receptor 7 (FZD7) is an oncogene involved in various malignancies. It is a receptor of Wingless (WNT) proteins. The protein possesses seven hydrophobic domains. The gene encoding FZD7 is localized on human chromosome 2q33.1.

Immunogen

FZD7 (NP_003498.1, 155 a.a. ~ 253 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GAGEICVGQNTSDGSGGPGGGPTAYPTAPYLPDLPFTALPPGASDGRGRPAFPFSCPRQLKVPPYLGYRFLGERDCGAPCEPGRANGLMYFKEEERRFA

Biochem/physiol Actions

Frizzled class receptor 7 (FZD7) modulates sensitivity to chemotherapeutic drugs. It functions in tumor initiation in melanoma cells. The protein acts as a mediator in canonical and non-canonical Wnt signaling pathways.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FZD7 drives in vitro aggressiveness in Stem-A subtype of ovarian cancer via regulation of non-canonical Wnt/PCP pathway
M Asad
Cell Death & Disease (2014)
Knockdown of frizzled-7 inhibits cell growth and metastasis and promotes chemosensitivity of esophageal squamous cell carcinoma cells by inhibiting Wnt signaling.
Liu X
Biochemical and Biophysical Research Communications (2017)
FRIZZLED7 Is Required for Tumor Inititation and Metastatic Growth of Melanoma Cells
Shweta T
PLoS ONE (2016)
Comparative integromics on FZD7 orthologs: conserved binding sites for PU. 1, SP1, CCAAT-box and TCF/LEF/SOX transcription factors within 5'-promoter region of mammalian FZD7 orthologs
Masuko Katoh
International Journal of Molecular Medicine (2007)
The Frizzled family: receptors for multiple signal transduction pathways
Hui-Chan Huang
Genome Biology (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service