Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1402396

Sigma-Aldrich

Monoclonal Anti-ZFP161 antibody produced in mouse

clone 4F7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC126126, ZBTB14, ZF5, ZNF478

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFP161(7541)

Immunogen

ZFP161 (NP_003400.2, 311 a.a. ~ 420 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TKGFTTQAHLKEHLKIHTGYKPYSCEVCGKSFIRAPDLKKHERVHSNERPFACHMCDKAFKHKSHLKDHERRHRGEKPFVCGSCTKAFAKASDLKRHENNMHSERKQVTP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia J Gorski et al.
Nucleic acids research, 39(22), 9536-9548 (2011-09-02)
A role for BRCA1 in the direct and indirect regulation of transcription is well established. However, a comprehensive view of the degree to which BRCA1 impacts transcriptional regulation on a genome-wide level has not been defined. We performed genome-wide expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service