Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1402382

Sigma-Aldrich

Monoclonal Anti-TPO antibody produced in mouse

clone 2A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MSA, TPX

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37.99 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TPO(7173)

General description

This gene encodes a membrane-bound glycoprotein. The encoded protein acts as an enzyme and plays a central role in thyroid gland function. The protein functions in the iodination of tyrosine residues in thyroglobulin and phenoxy-ester formation between pairs of iodinated tyrosines to generate the thyroid hormones, thyroxine and triiodothyronine. Mutations in this gene are associated with several disorders of thyroid hormonogenesis, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional splice variants have been described but their biological natures have not been determined. (provided by RefSeq)

Immunogen

TPO (NP_000538.3, 672 a.a. ~ 779 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
WENSHVFTDAQRRELEKHSLSRVICDNTGLTRVPMDAFQVGKFPEDFESCDSITGMNLEAWRETFPQDDKCGFPESVENGDFVHCEESGRRVLVYSCRHGYELQGREQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J F Stuijver et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 46(11), 789-793 (2014-04-03)
An excess of thyroid hormone leads to a prothrombotic state; however, the underlying pathophysiological mechanisms remain unknown. As evidence points towards an extensive "cross-talk" between the inflammatory and coagulation cascade, inflammation has been claimed as a possible mechanism through which
Mehmet Asik et al.
Endocrine, 46(2), 279-284 (2013-10-01)
We aimed to evaluate the prevalence of lactose intolerance (LI) in patients with Hashimoto's thyroiditis(HT) and the effects of lactose restriction on thyroid function in these patients. Eighty-three HT patients taking L-thyroxine (LT4) were enrolled, and lactose tolerance tests were
Kenji Iwaku et al.
Endocrine journal, 61(9), 875-881 (2014-07-11)
Following the accident at the Fukushima Daiichi Nuclear Power Station which occurred on March 11, 2011 due to the Eastern Japan Great Earthquake (the Accident), there have been concerns over elevation of the risk of thyroid cancer among children due
Neslihan Çuhaci et al.
Endocrine pathology, 25(3), 248-256 (2014-02-18)
Fine-needle aspiration biopsy (FNAB) has been widely accepted as the most accurate, safe, and cost-effective method for evaluation of thyroid nodules. The most challenging category in FNAB is atypia of undetermined significance (AUS) and follicular lesion of undetermined significance (FLUS).
Wenli Li et al.
The Journal of clinical endocrinology and metabolism, 99(9), E1602-E1609 (2014-06-07)
As a new antibody concept, natural bispecific antibodies (nBsAbs) have been detected in long-term passive immunization and some diseases, but their potential immunomodulatory role remains unclear. Hashimoto thyroiditis (HT) appears to fulfill the condition for nBsAb production but has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service